EMAIL THIS PAGE TO A FRIEND
Molecular and cellular biology
Loss of PTPN12 Stimulates Progression of ErbB2-Dependent Breast Cancer by Enhancing Cell Survival, Migration, and Epithelial-to-Mesenchymal Transition.
PMID 26391955
Abstract
PTPN12 is a cytoplasmic protein tyrosine phosphatase (PTP) reported to be a tumor suppressor in breast cancer, through its capacity to dephosphorylate oncogenic receptor protein tyrosine kinases (PTKs), such as ErbB2. However, the precise molecular and cellular impact of PTPN12 deficiency in breast cancer progression remains to be fully clarified. Here, we addressed this issue by examining the effect of PTPN12 deficiency on breast cancer progression in vivo, in a mouse model of ErbB2-dependent breast cancer using a conditional PTPN12-deficient mouse. Our studies showed that lack of PTPN12 in breast epithelial cells accelerated breast cancer development and lung metastases in vivo. PTPN12-deficient breast cancer cells displayed enhanced tyrosine phosphorylation of the adaptor Cas, the adaptor paxillin, and the kinase Pyk2. They exhibited no detectable increase in ErbB2 tyrosine phosphorylation. PTPN12-deficient cells were more resistant to anoikis and had augmented migratory and invasive properties. Enhanced migration was corrected by inhibiting Pyk2. PTPN12-deficient breast cancer cells also acquired partial features of epithelial-to-mesenchymal transition (EMT), a feature of more aggressive forms of breast cancer. Hence, loss of PTPN12 promoted tumor progression in a mouse model of breast cancer, supporting the notion that PTPN12 is a tumor suppressor in human breast cancer. This function was related to the ability of PTPN12 to suppress cell survival, migration, invasiveness, and EMT and to inhibit tyrosine phosphorylation of Cas, Pyk2, and paxillin. These findings enhance our understanding of the role and mechanism of action of PTPN12 in the control of breast cancer progression.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4500797
Anti-AKT antibody produced in rabbit, affinity isolated antibody
SAB4500799
Anti-AKT antibody produced in rabbit, affinity isolated antibody
SAB4500800
Anti-AKT antibody produced in rabbit, affinity isolated antibody
SAB4500802
Anti-AKT antibody produced in rabbit, affinity isolated antibody
SAB4500803
Anti-AKT antibody produced in rabbit, affinity isolated antibody
SAB4500796
Anti-AKT, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4500798
Anti-AKT, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4500004
Anti-AKT, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4300335
Anti-AKT1 (Ab-308) antibody produced in rabbit, affinity isolated antibody
SAB4300575
Anti-AKT1 (Ab-450) antibody produced in rabbit, affinity isolated antibody
SAB4300334
Anti-AKT1 (Ab-473) antibody produced in rabbit, affinity isolated antibody
GW22757 Anti-AKT1 (ab1) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
GW22775 Anti-AKT1 (ab2) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB2501934
Anti-AKT1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3500216
Anti-Akt1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA002891
Anti-AKT1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV06008 Anti-AKT1 antibody produced in rabbit, affinity isolated antibody
SAB4500007
Anti-AKT1 antibody produced in rabbit, affinity isolated antibody
SAB1406846
Anti-BCAR1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1401483
Anti-BCAR1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA042282
Anti-BCAR1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4503294
Anti-CASP3 (p17, Cleaved-Asp175) antibody produced in rabbit, affinity isolated antibody
SAB2501669
Anti-CASP3 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
AV00021
Anti-CASP3 antibody produced in rabbit, IgG fraction of antiserum
SAB4503291
Anti-CASP3 antibody produced in rabbit, affinity isolated antibody
SAB4300621
Anti-CDH2 antibody produced in rabbit, affinity isolated antibody
SAB4500002
Anti-CDH2, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB2701766
Anti-cse1l antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2100488
Anti-CSE1L antibody produced in rabbit, affinity isolated antibody
SAB1406701
Anti-EIF3G antibody produced in mouse, purified immunoglobulin, buffered aqueous solution