EMAIL THIS PAGE TO A FRIEND
Translational psychiatry
Shared functional defect in IP₃R-mediated calcium signaling in diverse monogenic autism syndromes.
PMID 26393489
Abstract
Autism spectrum disorder (ASD) affects 2% of children, and is characterized by impaired social and communication skills together with repetitive, stereotypic behavior. The pathophysiology of ASD is complex due to genetic and environmental heterogeneity, complicating the development of therapies and making diagnosis challenging. Growing genetic evidence supports a role of disrupted Ca(2+) signaling in ASD. Here, we report that patient-derived fibroblasts from three monogenic models of ASD-fragile X and tuberous sclerosis TSC1 and TSC2 syndromes-display depressed Ca(2+) release through inositol trisphosphate receptors (IP3Rs). This was apparent in Ca(2+) signals evoked by G protein-coupled receptors and by photoreleased IP3 at the levels of both global and local elementary Ca(2+) events, suggesting fundamental defects in IP3R channel activity in ASD. Given the ubiquitous involvement of IP3R-mediated Ca(2+) signaling in neuronal excitability, synaptic plasticity, gene expression and neurodevelopment, we propose dysregulated IP3R signaling as a nexus where genes altered in ASD converge to exert their deleterious effect. These findings highlight potential pharmaceutical targets, and identify Ca(2+) screening in skin fibroblasts as a promising technique for early detection of individuals susceptible to ASD.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4301087
Anti-ITPR2 antibody produced in rabbit, affinity isolated antibody