EMAIL THIS PAGE TO A FRIEND
British journal of haematology
Nelarabine, cyclosphosphamide and etoposide for adults with relapsed T-cell acute lymphoblastic leukaemia and lymphoma.
PMID 26403537
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML1736
Nelarabine, ≥98% (HPLC)
C11H15N5O5