EMAIL THIS PAGE TO A FRIEND
Genome announcements
Draft Genome Sequence of Bacillus sp. FJAT-27238 for Setting up Phylogenomic Analysis of Genomic Taxonomy of Bacillus-Like Bacteria.
PMID 26404591
Abstract
Bacillus sp. FJAT-27238 is a Gram-positive, spore-forming, and aerobic bacterium. Here, we report the draft genome sequence of Bacillus sp. FJAT-27238, with 6,134,829xa0bp, which will provide useful information for setting up phylogenomic analysis of the genomic taxonomy of Bacillus-like bacteria as well as for the functional gene mining and application of strain FJAT-27238. The genomic DNA G+C content was 47.37%.