EMAIL THIS PAGE TO A FRIEND
Oncoimmunology
Antigen targeting to dendritic cells combined with transient regulatory T cell inhibition results in long-term tumor regression.
PMID 26405564
Abstract
Therapeutic vaccinations against cancer are still largely ineffective. Major caveats are inefficient delivery of tumor antigens to dendritic cells (DCs) and excessive immune suppression by Foxp3(+) regulatory T cells (Tregs), resulting in defective T cell priming and failure to induce tumor regression. To circumvent these problems we evaluated a novel combinatorial therapeutic strategy. We show that tumor antigen targeting to DC-SIGN in humanized hSIGN mice via glycans or specific antibodies induces superior T cell priming. Next, this targeted therapy was combined with transient Foxp3(+) Treg depletion employing hSIGNxDEREG mice. While Treg depletion alone slightly delayed B16-OVA melanoma growth, only the combination therapy instigated long-term tumor regression in a substantial fraction of mice. This novel strategy resulted in optimal generation of antigen-specific activated CD8(+) T cells which accumulated in regressing tumors. Notably, Treg depletion also allowed the local appearance of effector T cells specific for endogenous B16 antigens. This indicates that antitumor immune responses can be broadened by therapies aimed at controlling Tregs in tumor environments. Thus, transient inhibition of Treg-mediated immune suppression potentiates DC targeted antigen vaccination and tumor-specific immunity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I9516
2-Propanol, for molecular biology, BioReagent, ≥99.5%
C3H8O
278475
2-Propanol, anhydrous, 99.5%
C3H8O
675431
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
C3H8O
437522
2-Propanol, ACS reagent, ≥99.5%
C3H8O
59304
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
C3H8O
40301
2-Propanol, semiconductor grade VLSI PURANAL (Honeywell 17604)
C3H8O
40219
2-Propanol, semiconductor grade PURANAL (Honeywell 17829)
C3H8O
40279
2-Propanol, semiconductor grade MOS PURANAL (Honeywell 17930)
C3H8O
270490
2-Propanol, 99.5%, HPLC grade
C3H8O
15-2360
2-Propanol, for HPLC
C3H8O
733458
2-Propanol, electronic grade, 99.999% trace metals basis
C3H8O
711438
4,6,6-Trimethyl-1,3,2-dioxaborinane solution, 1.0 M in dioxane
C6H13BO2
B7651
Brefeldin A, from Penicillium brefeldianum, ≥99% (HPLC and TLC)
C16H24O4
B6542
Brefeldin A, ≥99% (HPLC and TLC), BioXtra, for molecular biology
C16H24O4
B5936
Brefeldin A, from Penicillium brefeldianum, Ready Made Solution, 10 mg/mL in DMSO, 0.2 μm filtered
C16H24O4
E2028
Ethidium bromide monoazide, ≥95% (HPLC), solid
C21H18BrN5
W292907
Isopropyl alcohol, ≥99.7%, FCC, FG
C3H8O
79346
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
C36H56O8
P8139
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
C36H56O8