EMAIL THIS PAGE TO A FRIEND
Natural product research
Synthesis of deuterium-labelled substrates for the study of oleuropein biosynthesis in Olea europaea callus cultures.
PMID 26407195
Abstract
We propose the cell culture approach to investigate oleuropein (1) biogenesis in Olea europaea L. We suggest employing olive callus cultures to identify the iridoidic precursor of oleuropein. In fact, we confirmed that callus cells from olive shoot explants are able to produce key secoiridoid as 1. To enable this approach, we synthesised and characterised deuterium-labelled iridoidic precursors belonging both to the loganin and the 8-epiloganin series. These iridoids are [7,8-(2)H2]-7-deoxy-8-epi-loganin (2(D)), [8,10-(2)H2]-8-epi-loganin (4(D)) and [7,8-(2)H2]-7-deoxy-loganin (3(D)).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I6891
H-7
C14H17N3O2S