EMAIL THIS PAGE TO A FRIEND
Trends in ecology & evolution
Lamarckian Illusions.
PMID 26411613
Abstract
In recent years the term 'Lamarckian evolution' has become a household name for processes that do not follow classical Mendelian pattern of inheritance, and it is seen as a relevant complement to Darwinism. In this article I argue that bringing back Lamarck is unjustified and misleading.