EMAIL THIS PAGE TO A FRIEND
The international journal of cardiovascular imaging
Atrial fibrillation is associated with hematopoietic tissue activation and arterial inflammation.
PMID 26411879
Abstract
Inflammation is associated with the development of atrial fibrillation (AF). Activity in hematopoietic tissues, which produce inflammatory leukocytes, is closely related to systemic inflammation, arterial inflammation and cardiovascular events, but its relationship to AF is unknown. Using 18F-fluorodeoxyglucose (FDG) positron emission tomography (PET) imaging, we examined the relationships between AF, splenic metabolic activity and vascular inflammation. We conducted a cross sectional study of 70 participants: 35 with AF, who were matched (by age, sex and history of active cancer) to 35 controls without AF. Splenic metabolic activity and vascular aortic inflammation were measured by the mean FDG maximum standard uptake values (SUV Max) by PET. We examined (1) the association between AF and splenic activity, and (2) AF and aortic inflammation. The mean age of the population was 68.13 (standard deviation (SD) 8.98) years and 46 (65 %) participants were male. Splenic activity was higher in AF participants [2.31 (SD 0.45) vs. 2.07 (SD 0.37), p = 0.024], and remained significant after adjusting for demographic and clinical covariates. Aortic inflammation was also higher in AF participants [2.22 (SD 0.44) vs. 1.91 (SD 0.44), p = 0.004], and remained significant on multivariable analysis. Aortic inflammation and splenic activity were highly correlated (Pearson R = 0.61, p < 0.001). Atrial fibrillation is associated with higher hematopoietic tissue activation and arterial inflammation. Further studies are needed to clarify the mechanisms by which this cardio-splenic axis is implicated in AF.