EMAIL THIS PAGE TO A FRIEND
The Journal of clinical investigation
Dual-Affinity Re-Targeting proteins direct T cell-mediated cytolysis of latently HIV-infected cells.
PMID 26413868
Abstract
Enhancement of HIV-specific immunity is likely required to eliminate latent HIV infection. Here, we have developed an immunotherapeutic modality aimed to improve T cell-mediated clearance of HIV-1-infected cells. Specifically, we employed Dual-Affinity Re-Targeting (DART) proteins, which are bispecific, antibody-based molecules that can bind 2 distinct cell-surface molecules simultaneously. We designed DARTs with a monovalent HIV-1 envelope-binding (Env-binding) arm that was derived from broadly binding, antibody-dependent cellular cytotoxicity-mediating antibodies known to bind to HIV-infected target cells coupled to a monovalent CD3 binding arm designed to engage cytolytic effector T cells (referred to as HIVxCD3 DARTs). Thus, these DARTs redirected polyclonal T cells to specifically engage with and kill Env-expressing cells, including CD4+ T cells infected with different HIV-1 subtypes, thereby obviating the requirement for HIV-specific immunity. Using lymphocytes from patients on suppressive antiretroviral therapy (ART), we demonstrated that DARTs mediate CD8+ T cell clearance of CD4+ T cells that are superinfected with the HIV-1 strain JR-CSF or infected with autologous reservoir viruses isolated from HIV-infected-patient resting CD4+ T cells. Moreover, DARTs mediated CD8+ T cell clearance of HIV from resting CD4+ T cell cultures following induction of latent virus expression. Combined with HIV latency reversing agents, HIVxCD3 DARTs have the potential to be effective immunotherapeutic agents to clear latent HIV-1 reservoirs in HIV-infected individuals.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2702075
Anti-AP3B1 (N-terminal) antibody produced in rabbit
SAB1409916
Anti-AP3B1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA038737 Anti-AP3B1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2106308
Anti-AP3B1 antibody produced in rabbit, affinity isolated antibody
SAB1306027 ANTI-CHK(N-TERMINAL) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1405616
Anti-CHKA antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV53583
Anti-CHKA antibody produced in rabbit, affinity isolated antibody
HPA024153
Anti-CHKA antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410167
Anti-CHKA antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1401074
Anti-CHKB antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA018797
Anti-CHKB antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1408560
Anti-CHKB antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4500225
Anti-CHKB, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB2501783 Anti-PREP antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2104775
Anti-PREP antibody produced in rabbit, affinity isolated antibody
HPA031388
Anti-PREP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA031389
Anti-PREP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA031390
Anti-PREP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2104515
Anti-PREP, (N-terminal) antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB1302037
ANTI-TAC4 (C-TERM) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
WH0008546M6
Monoclonal Anti-AP3B1 antibody produced in mouse, clone 3B4, purified immunoglobulin, buffered aqueous solution
SAB1401302
Monoclonal Anti-PREP antibody produced in mouse, clone 2D7, purified immunoglobulin, buffered aqueous solution