EMAIL THIS PAGE TO A FRIEND
Stem cells and development
Direct Comparison of Wharton's Jelly and Bone Marrow-Derived Mesenchymal Stromal Cells to Enhance Engraftment of Cord Blood CD34(+) Transplants.
PMID 26414086
Abstract
Cotransplantation of CD34(+) hematopoietic stem and progenitor cells (HSPCs) with mesenchymal stromal cells (MSCs) enhances HSPC engraftment. For these applications, MSCs are mostly obtained from bone marrow (BM). However, MSCs can also be isolated from the Wharton's jelly (WJ) of the human umbilical cord. This source, regarded to be a waste product, enables a relatively low-cost MSC acquisition without any burden to the donor. In this study, we evaluated the ability of WJ MSCs to enhance HSPC engraftment. First, we compared cultured human WJ MSCs with human BM-derived MSCs (BM MSCs) for in vitro marker expression, immunomodulatory capacity, and differentiation into three mesenchymal lineages. Although we confirmed that WJ MSCs have a more restricted differentiation capacity, both WJ MSCs and BM MSCs expressed similar levels of surface markers and exhibited similar immune inhibitory capacities. Most importantly, cotransplantation of either WJ MSCs or BM MSCs with CB CD34(+) cells into NOD SCID mice showed similar enhanced recovery of human platelets and CD45(+) cells in the peripheral blood and a 3-fold higher engraftment in the BM, blood, and spleen 6 weeks after transplantation when compared to transplantation of CD34(+) cells alone. Upon coincubation, both MSC sources increased the expression of adhesion molecules on CD34(+) cells, although stromal cell-derived factor-1 (SDF-1)-induced migration of CD34(+) cells remained unaltered. Interestingly, there was an increase in CFU-GEMM when CB CD34(+) cells were cultured on monolayers of WJ MSCs in the presence of exogenous thrombopoietin, and an increase in BFU-E when BM MSCs replaced WJ MSCs in such cultures. Our results suggest that WJ MSC is likely to be a practical alternative for BM MSC to enhance CB CD34(+) cell engraftment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2702075
Anti-AP3B1 (N-terminal) antibody produced in rabbit
SAB1409916
Anti-AP3B1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA038737 Anti-AP3B1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2106308
Anti-AP3B1 antibody produced in rabbit, affinity isolated antibody
SAB2501783 Anti-PREP antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2104775
Anti-PREP antibody produced in rabbit, affinity isolated antibody
HPA031388
Anti-PREP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA031389
Anti-PREP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA031390
Anti-PREP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2104515
Anti-PREP, (N-terminal) antibody produced in rabbit, affinity isolated antibody, lyophilized powder
WH0008546M6
Monoclonal Anti-AP3B1 antibody produced in mouse, clone 3B4, purified immunoglobulin, buffered aqueous solution
SAB1401302
Monoclonal Anti-PREP antibody produced in mouse, clone 2D7, purified immunoglobulin, buffered aqueous solution