EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
The BAP1/ASXL2 Histone H2A Deubiquitinase Complex Regulates Cell Proliferation and Is Disrupted in Cancer.
PMID 26416890
Abstract
The deubiquitinase (DUB) and tumor suppressor BAP1 catalyzes ubiquitin removal from histone H2A Lys-119 and coordinates cell proliferation, but how BAP1 partners modulate its function remains poorly understood. Here, we report that BAP1 forms two mutually exclusive complexes with the transcriptional regulators ASXL1 and ASXL2, which are necessary for maintaining proper protein levels of this DUB. Conversely, BAP1 is essential for maintaining ASXL2, but not ASXL1, protein stability. Notably, cancer-associated loss of BAP1 expression results in ASXL2 destabilization and hence loss of its function. ASXL1 and ASXL2 use their ASXM domains to interact with the C-terminal domain (CTD) of BAP1, and these interactions are required for ubiquitin binding and H2A deubiquitination. The deubiquitination-promoting effect of ASXM requires intramolecular interactions between catalytic and non-catalytic domains of BAP1, which generate a composite ubiquitin-binding interface (CUBI). Notably, the CUBI engages multiple interactions with ubiquitin involving (i) the ubiquitin carboxyl hydrolase catalytic domain of BAP1, which interacts with the hydrophobic patch of ubiquitin, and (ii) the CTD domain, which interacts with a charged patch of ubiquitin. Significantly, we identified cancer-associated mutations of BAP1 that disrupt the CUBI and notably an in-frame deletion in the CTD that inhibits its interaction with ASXL1/2 and DUB activity and deregulates cell proliferation. Moreover, we demonstrated that BAP1 interaction with ASXL2 regulates cell senescence and that ASXL2 cancer-associated mutations disrupt BAP1 DUB activity. Thus, inactivation of the BAP1/ASXL2 axis might contribute to cancer development.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA017998
Anti-AP5Z1 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1407639
Anti-ASXL2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2106066
Anti-ASXL2 antibody produced in rabbit, affinity isolated antibody
SAB1406654
Anti-BAP1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
GW21184P Anti-c-Myc (ab1) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
GW21222 Anti-c-Myc (ab2) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
HPA050114
Anti-CDC6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1411704
Anti-CDC6 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4503728
Anti-CDC6, C-Terminal antibody produced in rabbit, affinity isolated antibody
GW21185 Anti-CDKN2A antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB2502016
Anti-Cdkn2a antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2501929
Anti-CDKN2A antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA035775
Anti-D4S234E antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2105613
Anti-D4S234E, (N-terminal) antibody produced in rabbit, affinity isolated antibody
HPA018864
Anti-FOXK1 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV47522
Anti-FOXK1 antibody produced in rabbit, affinity isolated antibody
AV37627
Anti-FOXK1 antibody produced in rabbit, IgG fraction of antiserum
SAB2105425
Anti-FOXK1 antibody produced in rabbit, affinity isolated antibody
SAB4500876
Anti-FOXK1, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB2100846
Anti-FOXK2 (ab1) antibody produced in rabbit, affinity isolated antibody
AV34775
Anti-FOXK2 (AB1) antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB2100847
Anti-FOXK2 (ab2) antibody produced in rabbit, affinity isolated antibody
AV100774
Anti-FOXK2 (AB2) antibody produced in rabbit, IgG fraction of antiserum
HPA027523
Anti-FOXK2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2103469
Anti-FOXK2, (C-terminal) antibody produced in rabbit, affinity isolated antibody
G7781
Anti-Glutathione-S-Transferase (GST) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
AV38600
Anti-HCFC1 antibody produced in rabbit, IgG fraction of antiserum
HPA018312
Anti-HCFC1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2104420
Anti-HCFC1 antibody produced in rabbit, affinity isolated antibody
AV51481
Anti-HIST2H2AA3 antibody produced in rabbit, IgG fraction of antiserum