EMAIL THIS PAGE TO A FRIEND
Oncotarget
Cytotoxic T cell responses are enhanced by antigen design involving the presentation of MUC1 peptide on cholera toxin B subunit.
PMID 26417929
Abstract
Induction of cytotoxic T lymphocytes (CTL) is critical to cancer vaccine based immunotherapy. Efforts to elicit CTLs against tumor MUC1 with peptide based vaccine have not been successful in clinical application. We have design a MUC1 vaccine by replacing B cell epitope of CTB with MUC1 VNTR peptide. Immunization with hybrid CTB-MUC1 plus aluminum hydroxide and CpG adujuvant (CTB-MUC1-Alum-CpG) induce MUC1-specific CTLs in mice. Moreover, this vaccination can prevent tumor growth and reduce tumor burden in MUC1+B16 mice model. Meanwhile, CTB-MUC1-Alum-CpG vaccination can promote Th1 cells and CD8+ T cells inflate to tumor tissue. Our approach might be applicable to other cancer vaccine design.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M2786
3-Maleimidobenzoic acid N-hydroxysuccinimide ester, crystalline
C15H10N2O6