EMAIL THIS PAGE TO A FRIEND
Oncotarget
TTK activates Akt and promotes proliferation and migration of hepatocellular carcinoma cells.
PMID 26418879
Abstract
Hepatocellular carcinoma (HCC) is one of the most malignant cancers with poor clinical outcome. The protein kinase human monopolar spindle 1 (hMps1/TTK) gene expression is significantly increased in HCCs. However, its contributions to hepatocarcinogenesis remain unclear. In this study, we found that TTK was overexpressed in 77.63% (118/152) HCC specimens. Elevated TTK expression positively correlated with large tumor size and presence of the portal vein tumor thrombus (PVTT). Demethylation in its promoter increased TTK expression in HCC. In vitro assays revealed that TTK not only promoted cell proliferation and anchorage-independent growth, but also cell migration. Subsequent investigations revealed that TTK activated Akt/mTOR pathway in a p53 dependent manner. We also found that TTK specific kinase inhibitor AZ3146 could decrease HCC cell growth. In conclusion, TTK contributes to HCC tumorigenesis via promoting cell proliferation and migration. It may serve as a novel biomarker and a potential target in HCC cancer therapy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4500797
Anti-AKT antibody produced in rabbit, affinity isolated antibody
SAB4500799
Anti-AKT antibody produced in rabbit, affinity isolated antibody
SAB4500800
Anti-AKT antibody produced in rabbit, affinity isolated antibody
SAB4500802
Anti-AKT antibody produced in rabbit, affinity isolated antibody
SAB4500803
Anti-AKT antibody produced in rabbit, affinity isolated antibody
SAB4500796
Anti-AKT, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4500798
Anti-AKT, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4500004
Anti-AKT, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4300335
Anti-AKT1 (Ab-308) antibody produced in rabbit, affinity isolated antibody
SAB4300575
Anti-AKT1 (Ab-450) antibody produced in rabbit, affinity isolated antibody
SAB4300334
Anti-AKT1 (Ab-473) antibody produced in rabbit, affinity isolated antibody
GW22757 Anti-AKT1 (ab1) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
GW22775 Anti-AKT1 (ab2) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB2501934
Anti-AKT1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3500216
Anti-Akt1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA002891
Anti-AKT1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV06008 Anti-AKT1 antibody produced in rabbit, affinity isolated antibody
SAB4500007
Anti-AKT1 antibody produced in rabbit, affinity isolated antibody
SAB4300601
Anti-MDM2 (Ab-166) antibody produced in rabbit, affinity isolated antibody
SAB2500625
Anti-MDM2 (Isoform) antibody produced in goat, affinity isolated antibody, buffered aqueous solution
GW21160 Anti-MDM2 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB4501849
Anti-MDM2 antibody produced in rabbit, affinity isolated antibody
SAB4501850
Anti-MDM2 antibody produced in rabbit, affinity isolated antibody
SAB4501851
Anti-MDM2, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB1306667
ANTI-P53 (S15) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1306713
ANTI-P53 (S315) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1306623
ANTI-P53 (SUMOYLATION SITE SPECIFIC) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1306675
ANTI-P53 (T55) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4503015
Anti-p53 antibody produced in rabbit, affinity isolated antibody
P4235 Anti-p53 antibody produced in sheep, fractionated antiserum, buffered aqueous solution