EMAIL THIS PAGE TO A FRIEND
Nature communications
The structural basis of Miranda-mediated Staufen localization during Drosophila neuroblast asymmetric division.
PMID 26423004
Abstract
During the asymmetric division of Drosophila neuroblasts (NBs), the scaffold Miranda (Mira) coordinates the subcellular distribution of cell-fate determinants including Staufen (Stau) and segregates them into the ganglion mother cells (GMCs). Here we show the fifth double-stranded RNA (dsRNA)-binding domain (dsRBD5) of Stau is necessary and sufficient for binding to a coiled-coil region of Mira cargo-binding domain (CBD). The crystal structure of Mira514-595/Stau dsRBD5 complex illustrates that Mira forms an elongated parallel coiled-coil dimer, and two dsRBD5 symmetrically bind to the Mira dimer through their exposed β-sheet faces, revealing a previously unrecognized protein interaction mode for dsRBDs. We further demonstrate that the Mira-Stau dsRBD5 interaction is responsible for the asymmetric localization of Stau during Drosophila NB asymmetric divisions. Finally, we find the CBD-mediated dimer assembly is likely a common requirement for Mira to recognize and translocate other cargos including brain tumour (Brat).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P0123
Anti-Paraoxonase 1 (PON1) antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB2501238
Anti-PON1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA001610
Anti-PON1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV41401
Anti-PON1 antibody produced in rabbit, affinity isolated antibody
SAB2104707
Anti-PON1 antibody produced in rabbit, affinity isolated antibody
SAB2102321
Anti-STAU1 (ab1) antibody produced in rabbit, affinity isolated antibody
SAB1406490
Anti-STAU1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1412071
ANTI-STAU1 antibody produced in mouse, clone 3C12, purified immunoglobulin, buffered aqueous solution
SAB1412648
ANTI-STAU1 antibody produced in mouse, clone 4D6, purified immunoglobulin, buffered aqueous solution
AV40536
Anti-STAU1 antibody produced in rabbit, affinity isolated antibody
AV40537
Anti-STAU1 antibody produced in rabbit, IgG fraction of antiserum
SAB3500565
Anti-STAU1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1402317
Monoclonal Anti-PON1, (C-terminal) antibody produced in mouse, clone 2H7, purified immunoglobulin, buffered aqueous solution
WH0006780M1
Monoclonal Anti-STAU1 antibody produced in mouse, clone 3D8, purified immunoglobulin, buffered aqueous solution
SAB1404419
Monoclonal Anti-STAU1, (C-terminal) antibody produced in mouse, clone 1E7, purified immunoglobulin, buffered aqueous solution