EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Amination of Aryl Halides and Esters Using Intensified Continuous Flow Processing.
PMID 26426000
Abstract
Significant process intensification of the amination reactions of aryl halides and esters has been demonstrated using continuous flow processing. Using this technology traditionally difficult amination reactions have been performed safely at elevated temperatures. These reactions were successfully conducted on laboratory scale coil reactor modules with 1 mm internal diameter (ID) and on a preparatory scale tubular reactor with 6 mm ID containing static mixers.