EMAIL THIS PAGE TO A FRIEND
Antonie van Leeuwenhoek
Phytomonospora cypria sp. nov., isolated from soil.
PMID 26427856
Abstract
A Gram-stain positive actinobacterial strain, designated KT1403(T), was isolated from a soil sample, collected from Karpaz, Magusa, Northern Cyprus, and characterised using a polyphasic approach. Morphological characteristics and chemotaxonomic data indicated that the strain belongs to the genus Phytomonospora. The cell wall of the novel strain contained meso-diaminopimelic acid and galactose, glucose and mannose as the major sugars in whole cell hydrolysates. The polar lipids in the cell membrane were identified as diphosphatidylglycerol, phosphatidylglycerol, phosphatidylethanolamine, phosphatidylinositol, an unidentified aminophospholipid and three unidentified glycolipids. The predominant menaquinones were MK-10(H6) and MK-10(H4). The major fatty acids were found to be iso C15:0 , anteiso C15:0 and anteiso C17:0. Phylogenetic analysis based on 16S rRNA gene sequences showed that strain KT1403(T) belongs to the genus Phytomonospora with a sequence similarity of 99.73xa0% with Phytomonospora endophytica, the type species of the genus. DNA-DNA hybridization further differentiated strain KT1403(T) from its near phylogenetic neighbour, P. endophytica DSM 45386(T) (29.0xa0±xa02.2xa0% DNA relatedness). Therefore, it is proposed that strain KT1403(T) represents a novel species of the genus Phytomonospora, for which the name Phytomonospora cypria sp. nov. is proposed. The type strain is KT1403(T) (=KCTC 29479(T)xa0=xa0DSM 46767(T)).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C7956 Coenzyme Q1, ≥95%
C14H18O4