EMAIL THIS PAGE TO A FRIEND
The American journal of cardiology
Effect of B-type natriuretic peptides on long-term outcomes after transcatheter aortic valve implantation.
PMID 26428025
Abstract
B-type natriuretic peptide (BNP) levels are elevated in patients with aortic stenosis (AS) and decrease acutely after replacement of the stenotic valve. The long-term prognostic value of BNP after transcatheter aortic valve implantation (TAVI) and the relative prognostic utility of single versus serial peri-interventional measurements of BNP and N-terminal prohormone BNP (NT-pro-BNP) are unknown. This study sought to determine the impact of BNP levels on long-term outcomes after TAVI and to compare the utility of BNP versus NT-pro-BNP measured before and after intervention. We analyzed 340 patients with severe AS and baseline pre-TAVI assessment of BNP. In 219 patients, BNP and NT-pro-BNP were measured serially before and after intervention. Clinical outcomes over 2 years were recorded. Patients with high baseline BNP (higher tertile ≥591 pg/ml) had increased risk of all-cause mortality (adjusted hazard ratio 3.16, 95% confidence interval 1.84 to 5.42; p <0.001) and cardiovascular death at 2 years (adjusted hazard ratio 3.37, 95% confidence interval 1.78 to 6.39; p <0.001). Outcomes were most unfavorable in patients with persistently high BNP before and after intervention. Comparing the 2 biomarkers, NT-pro-BNP levels measured after TAVI showed the highest prognostic discrimination for 2-year mortality (area under the curve 0.75; p <0.01). Baseline-to-discharge reduction, but not baseline levels of BNP, was related to New York Heart Association functional improvement. In conclusion, high preintervention BNP independently predicts 2-year outcomes after TAVI, particularly when elevated levels persist after the intervention. BNP and NT-pro-BNP and their serial periprocedural changes provide complementary prognostic information for symptomatic improvement and survival.