EMAIL THIS PAGE TO A FRIEND
Carbohydrate polymers
Influence of tiopronin, captopril and levamisole therapeutics on the oxidative degradation of hyaluronan.
PMID 26428153
Abstract
The ability to protect hyaluronic acid (HA) from oxidative degradation by cupric ions and ascorbate (production of (•)OH and peroxy-type radicals) during acute phase joint inflammation has been investigated using the following drugs: tiopronin, captopril, and levamisole. Radical scavenging activity, i.e. the propensity for donation of electrons was assessed for the drugs by ABTS and DPPH assays. The kinetics of HA degradation have been measured in the presence of each drug using rotational viscometry. The results of ABTS and DPPH assays show the highest radical scavenging activity for captopril, followed by tiopronin. For levamisole, no effect was observed. Captopril and tiopronin prevented HA degradation induced by (•)OH radicals in a similar manner, while tiopronin was more effective in scavenging peroxy-type radicals. On the other hand, levamisole was shown to be a pro-oxidant. Recovered HA fragments were characterized using FT-IR analysis, the incorporation of a sulphur atom from captopril and tiopronin but not from levamisole into the HA molecule was demonstrated.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L9756
(−)-Tetramisole hydrochloride, ≥99% (GC)
C11H12N2S · HCl
281689
1,1-Diphenyl-2-picrylhydrazine, 97%
C18H13N5O6
D9132
2,2-Diphenyl-1-picrylhydrazyl
C18H12N5O6
C4042
Captopril, ≥98% (HPLC), powder
C9H15NO3S
C8856
Captopril, meets USP testing specifications
C9H15NO3S
A5960
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
C6H8O6
A7506
L-Ascorbic acid, reagent grade, crystalline
C6H8O6
A0278
L-Ascorbic acid, reagent grade
C6H8O6
A4403
L-Ascorbic acid, powder, cell culture tested, γ-irradiated
C6H8O6
A4544
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
C6H8O6
A2218
L-Ascorbic acid, meets USP testing specifications
C6H8O6
A2174
L-Ascorbic acid, plant cell culture tested
C6H8O6
A2343
L-Ascorbic acid, meets USP testing specifications
C6H8O6
95209
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
C6H8O6
33034
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.7-100.5% (oxidimetric)
C6H8O6
95210
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
C6H8O6
47863
L-Ascorbic acid, analytical standard
C6H8O6
255564
L-Ascorbic acid, ACS reagent, ≥99%
C6H8O6
A92902
L-Ascorbic acid, 99%
C6H8O6
W210901
L-Ascorbic acid, FCC, FG
C6H8O6
M6635
N-(2-Mercaptopropionyl)glycine
C5H9NO3S
P5592
Potassium peroxodisulfate, BioXtra, ≥99.0%
K2O8S2
379824
Potassium persulfate, 99.99% trace metals basis
K2O8S2
216224
Potassium persulfate, ACS reagent, ≥99.0%
K2O8S2