EMAIL THIS PAGE TO A FRIEND
Oncotarget
Identification of thyroid tumor cell vulnerabilities through a siRNA-based functional screening.
PMID 26431489
Abstract
The incidence of thyroid carcinoma is rapidly increasing. Although generally associated with good prognosis, a fraction of thyroid tumors are not cured by standard therapy and progress to aggressive forms for which no effective treatments are currently available. In order to identify novel therapeutic targets for thyroid carcinoma, we focused on the discovery of genes essential for sustaining the oncogenic phenotype of thyroid tumor cells, but not required to the same degree for the viability of normal cells (non-oncogene addiction paradigm). We screened a siRNA oligonucleotide library targeting the human druggable genome in thyroid cancer BCPAP cell line in comparison with immortalized normal human thyrocytes (Nthy-ori 3-1). We identified a panel of hit genes whose silencing interferes with the growth of tumor cells, while sparing that of normal ones. Further analysis of three selected hit genes, namely Cyclin D1, MASTL and COPZ1, showed that they represent common vulnerabilities for thyroid tumor cells, as their inhibition reduced the viability of several thyroid tumor cell lines, regardless the histotype or oncogenic lesion. This work identified non-oncogenes essential for sustaining the phenotype of thyroid tumor cells, but not of normal cells, thus suggesting that they might represent promising targets for new therapeutic strategies.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1307257
ANTI-COPE(C-TERMINAL) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1101758
Anti-COPZ1 (31-45) antibody produced in rabbit, IgG fraction of antiserum
SAB4500348
Anti-Histone H3 (Acetyl-Lys14) antibody produced in rabbit, affinity isolated antibody
SAB4500349
Anti-Histone H3 (Acetyl-Lys18) antibody produced in rabbit, affinity isolated antibody
SAB4500350
Anti-Histone H3 (Acetyl-Lys23) antibody produced in rabbit, affinity isolated antibody
SAB4500351
Anti-Histone H3 (Acetyl-Lys27) antibody produced in rabbit, affinity isolated antibody
SAB4500347
Anti-Histone H3 (Acetyl-Lys9) antibody produced in rabbit, affinity isolated antibody
H9289
Anti-Histone H3 (N-terminal) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
H0164
Anti-Histone H3 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1410441
Anti-LYPLA3 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2104031
Anti-MAP4K5 antibody produced in rabbit, affinity isolated antibody
HPA036696
Anti-MAP4K5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA027175
Anti-MASTL antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4503392
Anti-MASTL, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB1401599
Anti-PLA2G15 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA041702
Anti-PLA2G15 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA041727
Anti-PLA2G15 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA002131
Anti-VCL antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
V4139 Anti-Vinculin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4503069
Anti-Vinculin antibody produced in rabbit, affinity isolated antibody
V9264
Anti-Vinculin antibody, Mouse monoclonal, clone hVIN-1, purified from hybridoma cell culture
EHU107331 MISSION® esiRNA, esiRNA human PSMC3 (esiRNA1)
EHU076481 MISSION® esiRNA, esiRNA human MASTL (esiRNA1)
EMU050781 MISSION® esiRNA, esiRNA targeting mouse Psmc3 (esiRNA1)
EMU075221 MISSION® esiRNA, esiRNA targeting mouse Mastl (esiRNA1)
WH0023659M1
Monoclonal Anti-LYPLA3 antibody produced in mouse, clone 3B11, purified immunoglobulin, buffered aqueous solution
WH0011183M1
Monoclonal Anti-MAP4K5 antibody produced in mouse, clone 3H3, purified immunoglobulin, buffered aqueous solution
WH0084930M1
Monoclonal Anti-MASTL antibody produced in mouse, clone 4B4-1B11, purified immunoglobulin, buffered aqueous solution
SAB1404522
Monoclonal Anti-VCL antibody produced in mouse, clone 3F8-1D4, purified immunoglobulin, buffered aqueous solution
SAB4200080
Monoclonal Anti-Vinculin antibody produced in mouse, clone V284, purified immunoglobulin, buffered aqueous solution