EMAIL THIS PAGE TO A FRIEND
Clinical immunology (Orlando, Fla.)
Activation of LXR attenuates collagen-induced arthritis via suppressing BLyS production.
PMID 26431776
Abstract
B-lymphocyte stimulator (BLyS) plays a critical role in the pathogenesis and progression of rheumatoid arthritis (RA). Liver X receptor (LXR), a nuclear receptor, has an important anti-inflammatory effect. However, it is unclear whether the BLyS expression is regulated by LXR. In this study, we found that treatment with LXR agonist in collagen-induced arthritis (CIA) mice significantly attenuated arthritis progression, and markedly decreased BLyS production in serum and splenocytes as well as the production of serum IFNγ and TGFβ. Activation of LXR in B lymphocytes dramatically suppressed the basal and IFNγ/TGFβ-induced BLyS expression. Moreover, LXR agonist prominently suppressed the binding of NF-κB to BLyS promoter region, and decreased the promoter's transcriptional activity. Additionally, activation of LXR obviously repressed IFNγ-induced STAT1 activation and TGFβ-induced SMAD3 activation. These results indicated that downregulation of BLyS may be a novel mechanism by which LXR ameliorates RA, and LXR/BLyS pathway may serve as a novel target for the treatment of RA.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PRS2221
Anti-BAFF antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
B4805
Anti-BAFF antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB4504210
Anti-phospho-Smad3 (pSer204) antibody produced in rabbit, affinity isolated antibody
SAB4504211
Anti-phospho-Smad3 (pSer208) antibody produced in rabbit, affinity isolated antibody
SAB4504212
Anti-phospho-Smad3 (pSer213) antibody produced in rabbit, affinity isolated antibody
SAB4503781
Anti-phospho-Smad3 (pSer425) antibody produced in rabbit, affinity isolated antibody
SAB4300253
Anti-phospho-SMAD3 (pSer425) antibody produced in rabbit, affinity isolated antibody
SAB4504209
Anti-phospho-Smad3 (pThr179) antibody produced in rabbit, affinity isolated antibody
SAB4300133
Anti-phospho-STAT1 (pSer727) antibody produced in rabbit, affinity isolated antibody
SAB4504538
Anti-phospho-STAT1 (pSer727) antibody produced in rabbit, affinity isolated antibody
S2565 Anti-phospho-STAT1 (pTyr701) antibody produced in rabbit, affinity isolated antibody, aqueous glycerol solution
SAB4300032
Anti-phospho-STAT1 (pTyr701) antibody produced in rabbit, affinity isolated antibody
SAB4504539
Anti-phospho-STAT1 (pTyr701) antibody produced in rabbit, affinity isolated antibody
SAB4502868
Anti-STAT1, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4502869
Anti-STAT1, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB1407039
Anti-TNFSF13B antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2105077
Anti-TNFSF13B, (N-terminal) antibody produced in rabbit, affinity isolated antibody
WH0010673M1
Monoclonal Anti-TNFSF13B antibody produced in mouse, clone 1C11, purified immunoglobulin, buffered aqueous solution
SAB1404858
Monoclonal Anti-TNFSF13B antibody produced in mouse, clone 2C11, purified immunoglobulin, buffered aqueous solution
SAB1408787
Monoclonal Anti-TNFSF13B antibody produced in mouse, clone 3G6, purified immunoglobulin, buffered aqueous solution