EMAIL THIS PAGE TO A FRIEND
The American journal of cardiology
Clinical Outcome and Cost Analysis of Sutureless Versus Transcatheter Aortic Valve Implantation With Propensity Score Matching Analysis.
PMID 26433277
Abstract
Surgical sutureless and interventional transcatheter aortic valve prostheses are nowadays extensively adopted in high-risk elderly patients. An explorative analysis was carried out to compare the clinical outcome and costs associated to these approaches. Since 2010, a total of 626 patients were distributed between transcatheter aortic valve implantation (TAVI; n = 364) and sutureless (n = 262) groups. Patients of both groups were not comparable for clinical and surgical characteristics, but many patients were in a "gray zone"; therefore, a retrospective propensity score analysis was possible and performed. For the matched pair samples, postoperative, follow-up clinical data, and costs data were obtained. In-hospital death occurred in 5 patients in sutureless group and 3 patients in TAVI group (p = 0.36). Blood transfusions were higher in sutureless group (2.1 ± 2.3 vs 0.4 ± 1.0 U). TAVI group had a shorter intensive care unit and hospital stay (2.2 ± 2.7 vs 3.2 ± 3.5 days, p = 0.037; 12 ± 6 vs 14 ± 6 days, p = 0.017). No differences in postoperative neurologic (p = 0.361), renal (p = 0.106), or respiratory (p = 0.391) complications were observed between groups. At follow-up (24.5 ± 13.8 months), 1 patient in sutureless group and 7 patients in TAVI group died (p = 0.032). Paravalvular leakage occurred more frequently in patients in TAVI group (35 [34%] vs 7 [6.9%]; p <0.001) with an impact on follow-up survival rate. The costs associated to the 2 procedures are similar when the cost of the device was excluded (p = 0.217). When included, the sutureless approach resulted a cost saving (€22,451 vs €33,877, p <0.001). In conclusion, the patients in the "gray zone" record a satisfying clinical outcome after sutureless surgery and TAVI. Patients in the sutureless group endure more hospital complications, but TAVI entails a higher follow-up mortality. On the costs aspects, TAVI technologies are more expensive, and it reflects on higher overall hospital costs.