EMAIL THIS PAGE TO A FRIEND
Macromolecular bioscience
Thiol-ene Photocrosslinking of Cytocompatible Resilin-Like Polypeptide-PEG Hydrogels.
PMID 26435299
Abstract
A range of chemical strategies have been used for crosslinking recombinant polypeptide hydrogels, although only a few have employed photocrosslinking approaches. Here, we capitalize on the novel insect protein, resilin, and the versatility of click reactions to introduce a resilin-like polypeptide (RLP) that is capable of photoinitiated thiol-ene crosslinking. Lysine residues of the RLP were functionalized with norbornene acid as confirmed via 1H-NMR spectroscopy. The RLPNs were subsequently photocrosslinked with multi-arm PEG thiols in the presence of a photoinitiator to form elastic hybrid hydrogels. The crosslinking reaction and resulting RLP-PEG networks demonstrated cytocompatibility with human mesenchymal stem cells in both 2D cell-adhesion and 3D photoencapsulation studies.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
718149
exo-5-Norbornenecarboxylic acid, 97%
C8H10O2