EMAIL THIS PAGE TO A FRIEND
The Journal of clinical investigation
Sialylation of IgG Fc domain impairs complement-dependent cytotoxicity.
PMID 26436649
Abstract
IgG molecules exert both pro- and antiinflammatory effector functions based on the composition of the fragment crystallizable (Fc) domain glycan. Sialylated IgG Fc domains have antiinflammatory properties that are attributed to their ability to increase the activation threshold of innate effector cells to immune complexes by stimulating the upregulation of the inhibitory Fcγ receptor IIB (FcγRIIB). Here, we report that IgG Fc sialylation of human monoclonal IgG1 molecules impairs their efficacy to induce complement-mediated cytotoxicity (CDC). Fc sialylation of a CD20-targeting antibody had no impact on antibody-dependent cellular cytotoxicity and did not change the affinity of the antibody for activating Fcγ receptors. In contrast, the presence of sialic acid abrogated the increased binding of C1q to Fc-galactosylated IgG1 and resulted in decreased levels of C3b deposition on the cell surface. Similar to monoclonal antibodies, sialic acid inhibited the increased C1q binding to galactosylated Fc fragments in human polyclonal IgG. In sera derived from patients with chronic inflammatory demyelinating polyneuropathy, an autoimmune disease of the peripheral nervous system in which humoral immune responses mediate tissue damage, induction of IgG Fc sialylation was associated with clinical disease remission. Thus, impairment of CDC represents an FcγR-independent mechanism by which Fc-sialylated glycovariants might limit proinflammatory IgG effector functions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A0170 Anti-Human IgG (Fc specific)−Peroxidase antibody produced in goat, affinity isolated antibody
M0820
Anti-MOG (C-terminal) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
M0695
Anti-MOG (N-terminal) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2500639
Anti-MOG antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1406138
Anti-MOG antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA021873
Anti-MOG antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410792
Anti-MOG antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB3701024 Anti-Mouse IgG (Fc specific)-Alkaline Phosphatase antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB3701025 Anti-Mouse IgG (Fc specific)-Biotin antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3701022 Anti-Mouse IgG (Fc specific)-Fluorescein antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3701023 Anti-Mouse IgG (Fc specific)-Peroxidase antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3701020 Anti-Mouse IgG (Fc specific)-Rhodamine antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3701026 Anti-Mouse IgG (Fc specific)-Texas Red® antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3701038 Anti-Mouse IgG (γ-chain specific)-Alkaline Phosphatase antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB3701039 Anti-Mouse IgG (γ-chain specific)-Biotin antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3701036 Anti-Mouse IgG (γ-chain specific)-Fluorescein antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3701037 Anti-Mouse IgG (γ-chain specific)-Peroxidase antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3701034 Anti-Mouse IgG (γ-chain specific)-Rhodamine antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3701040 Anti-Mouse IgG (γ-chain specific)-Texas Red® antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3701166 Anti-Mouse IgG (H+L), F(ab) fragment-Biotin antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3701164 Anti-Mouse IgG (H+L), F(ab) fragment-Fluorescein antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3701165 Anti-Mouse IgG (H+L), F(ab) fragment-Peroxidase antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3701162 Anti-Mouse IgG (H+L), F(ab) fragment-Rhodamine antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3701167 Anti-Mouse IgG (H+L), F(ab) fragment-Texas Red® antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB4600018
Anti-Mouse IgG (H+L), highly cross-adsorbed, CF 350 antibody produced in rabbit, ~2 mg/mL, affinity isolated antibody
SAB4600056
Anti-Mouse IgG (H+L), highly cross-adsorbed, CF 488A antibody produced in rabbit, ~2 mg/mL, affinity isolated antibody
SAB4600073
Anti-Mouse IgG (H+L), highly cross-adsorbed, CF 555 antibody produced in rabbit, ~2 mg/mL, affinity isolated antibody
SAB4600089
Anti-Mouse IgG (H+L), highly cross-adsorbed, CF 568 antibody produced in rabbit, ~2 mg/mL, affinity isolated antibody
SAB4600121
Anti-Mouse IgG (H+L), highly cross-adsorbed, CF 594 antibody produced in rabbit, ~2 mg/mL, affinity isolated antibody
SAB4600150
Anti-Mouse IgG (H+L), highly cross-adsorbed, CF 633 antibody produced in rabbit, ~2 mg/mL, affinity isolated antibody