EMAIL THIS PAGE TO A FRIEND
British journal of pharmacology
Quetiapine and its metabolite norquetiapine: translation from in vitro pharmacology to in vivo efficacy in rodent models.
PMID 26436896
Abstract
Quetiapine has a range of clinical activity distinct from other atypical antipsychotic drugs, demonstrating efficacy as monotherapy in bipolar depression, major depressive disorder and generalized anxiety disorder. The neuropharmacological mechanisms underlying this clinical profile are not completely understood; however, the major active metabolite, norquetiapine, has been shown to have a distinct in vitro pharmacological profile consistent with a broad therapeutic range and may contribute to the clinical profile of quetiapine. We evaluated quetiapine and norquetiapine, using in vitro binding and functional assays of targets known to be associated with antidepressant and anxiolytic drug actions and compared these activities with a representative range of established antipsychotics and antidepressants. To determine how the in vitro pharmacological properties translate into in vivo activity, we used preclinical animal models with translational relevance to established antidepressant-like and anxiolytic-like drug action. Norquetiapine had equivalent activity to established antidepressants at the noradrenaline transporter (NET), while quetiapine was inactive. Norquetiapine was active in the mouse forced swimming and rat learned helplessness tests. In in vivo receptor occupancy studies, norquetiapine had significant occupancy at NET at behaviourally relevant doses. Both quetiapine and norquetiapine were agonists at 5-HT1A receptors, and the anxiolytic-like activity of norquetiapine in rat punished responding was blocked by the 5-HT1A antagonist, WAY100635. Quetiapine and norquetiapine have multiple in vitro pharmacological actions, and results from preclinical studies suggest that activity at NET and 5-HT1A receptors contributes to the antidepressant and anxiolytic effects in patients treated with quetiapine.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
766208
Diphenylammonium Triflate, 97%
C13H12F3NO3S
N151
Nisoxetine hydrochloride, solid, ≥98% (HPLC)
C17H21NO2 · HCl