EMAIL THIS PAGE TO A FRIEND
Marine drugs
Mycemycins A-E, New Dibenzoxazepinones Isolated from Two Different Streptomycetes.
PMID 26437421
Abstract
Five new dibenzoxazepinone derivatives, mycemycins A-E (1-5), were isolated from the ethanol extracts of mycelia of two different streptomycetes. 1 and 2 were isolated from an acidic red soil-derived strain, Streptomyces sp. FXJ1.235, and 3-5 from a gntR gene-disrupted deep-sea strain named Streptomyces olivaceus FXJ8.012Δ1741. The structures of mycemycins were elucidated by a combination of spectroscopic analyses, including 1D- and 2D-NMR techniques.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
11468120910
Reverse Transcriptase Assay, colorimetric