EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
p21-activated Kinases (PAKs) Mediate the Phosphorylation of PREX2 Protein to Initiate Feedback Inhibition of Rac1 GTPase.
PMID 26438819
Abstract
Phosphatidylinositol 3,4,5-trisphosphate (PIP3)-dependent Rac exchanger 2 (PREX2) is a guanine nucleotide exchange factor (GEF) for the Ras-related C3 botulinum toxin substrate 1 (Rac1) GTPase, facilitating the exchange of GDP for GTP on Rac1. GTP-bound Rac1 then activates its downstream effectors, including p21-activated kinases (PAKs). PREX2 and Rac1 are frequently mutated in cancer and have key roles within the insulin-signaling pathway. Rac1 can be inactivated by multiple mechanisms; however, negative regulation by insulin is not well understood. Here, we show that in response to being activated after insulin stimulation, Rac1 initiates its own inactivation by decreasing PREX2 GEF activity. Following PREX2-mediated activation of Rac1 by the second messengers PIP3 or Gβγ, we found that PREX2 was phosphorylated through a PAK-dependent mechanism. PAK-mediated phosphorylation of PREX2 reduced GEF activity toward Rac1 by inhibiting PREX2 binding to PIP3 and Gβγ. Cell fractionation experiments also revealed that phosphorylation prevented PREX2 from localizing to the cellular membrane. Furthermore, the onset of insulin-induced phosphorylation of PREX2 was delayed compared with AKT. Altogether, we propose that second messengers activate the Rac1 signal, which sets in motion a cascade whereby PAKs phosphorylate and negatively regulate PREX2 to decrease Rac1 activation. This type of regulation would allow for transient activation of the PREX2-Rac1 signal and may be relevant in multiple physiological processes, including diseases such as diabetes and cancer when insulin signaling is chronically activated.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1409833
Anti-DEPDC2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA015234
Anti-PREX2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
A7345 Anti-V5 Agarose Affinity Gel antibody produced in mouse, purified immunoglobulin, clone V5-10
E0396
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
C14H24N2O10
E3889
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, for molecular biology, ≥97.0%
C14H24N2O10
E4378
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
C14H24N2O10
792527
Imidazole, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
C3H4N2
I5513
Imidazole, for molecular biology, ≥99% (titration)
C3H4N2
I0250
Imidazole, ≥99% (titration), crystalline
C3H4N2
I2399
Imidazole, ACS reagent, ≥99% (titration)
C3H4N2
56750
Imidazole, puriss. p.a., ≥99.5% (GC)
C3H4N2
I202
Imidazole, ReagentPlus®, 99%
C3H4N2
56748
Imidazole, BioUltra, for molecular biology, ≥99.5% (GC)
C3H4N2
56749
Imidazole, BioUltra, ≥99.5% (GC)
C3H4N2
68268
Imidazole buffer Solution, BioUltra, 1 M in H2O
C3H4N2
P0040
PP1, ≥98% (HPLC)
C16H19N5
S7920
Sodium fluoride, BioXtra, ≥99%
FNa
S6776
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
FNa
919
Sodium fluoride solution
FNa