EMAIL THIS PAGE TO A FRIEND
Nature communications
Acidic phospholipids govern the enhanced activation of IgG-B cell receptor.
PMID 26440273
Abstract
B cells that express the isotype-switched IgG-B cell receptor (IgG-BCR) are one of the driving forces for antibody memory. To allow for a rapid memory IgG antibody response, IgG-BCR evolved into a highly effective signalling machine. Here, we report that the positively charged cytoplasmic domain of mIgG (mIgG-tail) specifically interacts with negatively charged acidic phospholipids. The key immunoglobulin tail tyrosine (ITT) in mIgG-tail is thus sequestered in the membrane hydrophobic core in quiescent B cells. Pre-disruption of such interaction leads to excessive recruitment of BCRs and inflated BCR signalling upon antigen stimulation, resulting in hyperproliferation of primary B cells. Physiologically, membrane-sequestered mIgG-tail can be released by antigen engagement or Ca(2+) mobilization in the initiation of B cell activation. Our studies suggest a novel regulatory mechanism for how dynamic association of mIgG-tail with acidic phospholipids governs the enhanced activation of IgG-BCR.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
63371
2-Oleoyl-1-palmitoyl-sn-glycero-3-phospho-rac-(1-glycerol) sodium salt, ≥98.0% (TLC)
C40H76NaO10P
21888
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
C29H19NO11
87444
5-Carboxy-fluorescein diacetate N-succinimidyl ester, for fluorescence, ≥95.0% (HPLC)
C29H19NO11
SAB3701027 Anti-Mouse IgG (Fc specific), F(ab′)2 fragment, highly cross adsorbed antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3701127 Anti-Mouse IgG (H+L), F(ab′)2 fragment antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3701157 Anti-Mouse IgG (H+L), F(ab) fragment antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3700991 Anti-Mouse IgG F(ab′)2 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3701013 Anti-Mouse IgG F(ab′)2, F(ab′)2 fragment, highly cross absorbed antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB4300282
Anti-phospho-SYK (pTyr323) antibody produced in rabbit, affinity isolated antibody
SAB4503994
Anti-phospho-SYK (pTyr323) antibody produced in rabbit, affinity isolated antibody
SAB4503838
Anti-phospho-SYK (pTyr348) antibody produced in rabbit, affinity isolated antibody
SAB4503839
Anti-phospho-SYK (pTyr525) antibody produced in rabbit, affinity isolated antibody
SAB3700849 Anti-Rabbit IgG (Fc specific), highly cross adsorbed antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3700946 Anti-Rabbit IgG (H+L), F(ab′)2 fragment antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3700970 Anti-Rabbit IgG (H+L), F(ab) fragment antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3700840
Anti-Rabbit IgG F(ab′)2, F(ab′)2 fragment, highly cross absorbed antibody produced in goat, affinity isolated antibody, buffered aqueous solution
I6758
IPTG, ≥99% (TLC), ≤0.1% Dioxane
C9H18O5S
PHG0010
Isopropyl β-D-1-thiogalactopyranoside, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production.
C9H18O5S
I5502
Isopropyl β-D-1-thiogalactopyranoside, ≥99% (TLC)
C9H18O5S
I1284
Isopropyl β-D-thiogalactopyranoside solution, ReadyMade IPTG solution for Blue-white screening
C9H18O5S
R6626
Rhodamine B, ≥95% (HPLC)
C28H31ClN2O3
83689
Rhodamine B, for fluorescence
C28H31ClN2O3