EMAIL THIS PAGE TO A FRIEND
Chirality
Fast Screening of Whole Blood Samples and Pharmaceutical Compounds for Enantiorecognition of Free L-T3 , L-T4 , and D-T4.
PMID 26447904
Abstract
A fast screening method of whole blood was proposed for enantiorecognition of free L-T3 , L-T4 , and D-T4 . Stochastic microsensors based on four inulins (IN, IQ, TEX, and HD) immobilized on diamond paste (DP) were used for recognition of free L-T3 , L-T4 , and D-T4 . For the enantiorecognition of free L-T4 and D-T4 in whole blood and pharmaceutical samples, the best microsensor was the one based on TEX/DP (wide linear concentration ranges, and low limits of quantification). The best limit of detection for the assay of free L-T3 (400 fmol/L) was recorded using the microsensors based on HD/DP, while for the assay of free L-T4, and D-T4 the best limit of determination (1 pmol/L) was recorded using the TX/DP-based microsensor. For the enantiorecognition of free L-T3 in whole blood and pharmaceutical samples the best microsensor was the one based on HD/DP (the wider linear concentration range, and the lower limit of quantification - of pmol/L magnitude order). For the enantiorecognition of free L-T3 in whole blood and pharmaceutical samples the best microsensor was the one based on HD/DP (the wider linear concentration range, and the lower limit of quantification - of pmol/L magnitude order). Free L-T3 , L-T4 , and D-T4 were recovered with high reliabilities in whole blood samples (recoveries higher than 99.00%, with RSD values lower than 1.00%) and pharmaceutical samples (recoveries higher than 95.00% with RSD values lower than 1.00%).