EMAIL THIS PAGE TO A FRIEND
Oncoimmunology
Interleukin-17A promotes the growth of human germinal center derived non-Hodgkin B cell lymphoma.
PMID 26451300
Abstract
Interleukin (IL)-17A belongs to IL-17 superfamily and binds the heterodimeric IL-17 receptor (R)(IL-17RA/IL-17RC). IL-17A promotes germinal center (GC) formation in mouse models of autoimmune or infectious diseases, but the role of IL-17A/IL-17AR complex in human neoplastic GC is unknown. In this study, we investigated expression and function of IL-17A/IL-17AR in the microenvironments of 44 B cell non-Hodgkin lymphomas (B-NHL) of GC origin (15 follicular lymphomas, 17 diffuse large B cells lymphomas and 12 Burkitt lymphomas) and 12 human tonsil GC. Furthermore, we investigated the role of IL-17A in two in vivo models of GC B cell lymphoma, generated by s.c. injection of SU-DHL-4 and OCI-Ly8 cell lines in Severe combined immunodeficiency (SCID)/Non Obese Diabetic (NOD) mice. We found that: (i) B-NHL cell fractions and tonsil GC B cells expressed IL-17RA/IL-17RC, (ii) IL-17A signaled in both cell types through NF-kBp65, but not p38, ERK-1/2, Akt or NF-kBp50/105, phosphorylation, (iii) IL-17A was expressed in T cells and mast cells from neoplastic and normal GC microenvironments, (iv) IL-17A rendered tonsil GC B cells competent to migrate to CXCL12 and CXCL13 by downregulating RGS16 expression; (v) IL-17A stimulated in vitro proliferation of primary B-NHL cells; (vi) IL-17A (1xa0μg/mouse-per dose) stimulated B-NHL growth in two in vivo models by enhancing tumor cell proliferation and neo-angiogenesis. This latter effect depended on IL-17A-mediated induction of pro-angiogenic gene expression in tumor cells and direct stimulation of endothelial cells. These data define a previously unrecognized role of human IL-17A in promoting growth of GC-derived B-NHL and modulating normal GC B cell trafficking.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV47287
Anti-GDE1 antibody produced in rabbit, affinity isolated antibody
SAB1406012
Anti-IGHD antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1101232
Anti-IL17RA (166-180) antibody produced in rabbit, IgG fraction of antiserum
SAB2501167
Anti-Il17ra antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1401600
Anti-IL17RA antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1408127
Anti-IL17RC antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA019885 Anti-IL17RC antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2700878
Anti-IL17RC antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1407504
Anti-MIR16 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA000451
Anti-MKI67 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA001164
Anti-MKI67 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
GW21321 Anti-RGS16 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
AV34040
Anti-RGS16 antibody produced in rabbit, IgG fraction of antiserum
HPA053250
Anti-RGS16 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA055824
Anti-RGS16 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4502634
Anti-RGS16, C-Terminal antibody produced in rabbit, affinity isolated antibody
F3651
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
C21H11NO5S
46950
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
C21H11NO5S
F7250
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
C21H11NO5S
F4274
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
C21H11NO5S
46951
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
C21H11NO5S
I2768 Monoclonal Anti-IgD antibody produced in mouse, clone HJ9, purified immunoglobulin, buffered aqueous solution
WH0051573M4
Monoclonal Anti-MIR16 antibody produced in mouse, clone 2H6, purified immunoglobulin, buffered aqueous solution
WH0004288M1
Monoclonal Anti-MKI67 antibody produced in mouse, clone 7B8, purified immunoglobulin, buffered aqueous solution
AMAB90870
Monoclonal Anti-MKI67 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1234, purified immunoglobulin, buffered aqueous glycerol solution
SAB5300423 Monoclonal Anti-MKI67 antibody produced in mouse, clone 4A1, ascites fluid
SAB5300424 Monoclonal Anti-MKI67 antibody produced in mouse, clone 9C12B2, ascites fluid
SAB5300425 Monoclonal Anti-MKI67 antibody produced in mouse, clone 8D5, ascites fluid
SAB4100422
Monoclonal Anti-RGS16 antibody produced in mouse, clone RGS16-B350, culture supernatant