EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
In vitro studies of serum albumin interaction with poly(D,L-lactide) nanospheres loaded by hydrophobic cargo.
PMID 26452101
Abstract
The various polymer-based nanocarriers are very attractive for in vitro and in vivo bioapplications. A new type of a promising drug delivery systems for cancer tissues-poly(D,L-lactide) nanospheres stabilized with Cremophor EL and loaded with hydrophobic cyanines (IR-780 or ZnPc) or curcumin (CUR) were fabricated by the nanoprecipitation method. The Cremophor EL/PLA/water nanospheres demonstrated regular shape, low polydispersity (PdI<0.3) and high entrapment efficiency of selected cargo (over 90%). The size of those nanoconstructs below 130 nm are in the desired nanocarriers size range for tumor delivery. Low level of in vitro drug release from loaded nanospheres after long-time storage indicates their good stability. The half-life of nanocarriers in the circulation, and their biodistribution after parenteral administration are associated with the ability of plasma proteins adsorption. For these reasons the affinity of obtained nanospheres for albumin as a major plasma protein was in vitro investigated. The binding of nanocarrier containing cyanine IR-780 with albumin immobilized in the wells of polystyrene plate occurred with lower efficiency than analogs loaded with ZnPc or CUR. Similar relationships were observed after UV-vis spectra analysis of nanospheres in the presence of albumin at various protein concentrations.