EMAIL THIS PAGE TO A FRIEND
Journal of the American Veterinary Medical Association
Pharmacologic profile of methoprene, an insect growth regulator, in cattle, dogs, and cats.
PMID 2645260
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33375
Methoprene, PESTANAL®, analytical standard
C19H34O3