EMAIL THIS PAGE TO A FRIEND
Neurobiology of aging
Thrombospondin-1 prevents amyloid beta-mediated synaptic pathology in Alzheimer's disease.
PMID 26452999
Abstract
Alzheimer's disease (AD) is characterized by impaired cognitive function and memory loss, which are often the result of synaptic pathology. Thrombospondin (TSP) is an astrocyte-secreted protein, well known for its function as a modulator of synaptogenesis and neurogenesis. Here, we investigated the effects of TSP-1 on AD pathogenesis. We found that the level of TSP-1 expression was decreased in AD brains. When we treated astrocytes with amyloid beta (Aβ), secreted TSP-1 was decreased in autophagy-dependent manner. In addition, treatment with Aβ induced synaptic pathology, such as decreased dendritic spine density and reduced synaptic activity. These effects were prevented by coincubation of TSP-1 with Aβ, which acts through the TSP-1 receptor alpha-2-delta-1 in neurons. Finally, intrasubicular injection with TSP-1 into AD model mouse brains mitigated the Aβ-mediated reduction of synaptic proteins and related signaling pathways. These results indicate that TSP-1 is a potential therapeutic target in AD pathogenesis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4503258
Anti-Calnexin, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB1401057
Anti-CD47 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA044659
Anti-CD47 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4300405
Anti-GRIN1 (Ab-896) antibody produced in rabbit, affinity isolated antibody
SAB4300538
Anti-GRIN1 (Ab-897) antibody produced in rabbit, affinity isolated antibody
SAB2101002
Anti-GZMA antibody produced in rabbit, affinity isolated antibody
HPA024504
Anti-NDOR1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410533
Anti-NDOR1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4501301
Anti-NMDAR1, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4501302
Anti-NMDAR1, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4501303
Anti-NMDAR1, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4503908
Anti-phospho-CREB (pSer121) antibody produced in rabbit, affinity isolated antibody
SAB4504375
Anti-phospho-CREB (pSer133) antibody produced in rabbit, affinity isolated antibody
SAB4503757
Anti-phospho-CREB (pSer142) antibody produced in rabbit, affinity isolated antibody
SAB4504577
Anti-phospho-CREB (pThr100) antibody produced in rabbit, affinity isolated antibody
SAB1306301 ANTI-PHOSPHO-CREB(S133) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4300217
Anti-phospho-CREB1 (pSer129) antibody produced in rabbit, affinity isolated antibody
SAB4300040
Anti-phospho-CREB1 (pSer133) antibody produced in rabbit, affinity isolated antibody
SAB1401385
Anti-THBS1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SRP6055
Granzyme A human, recombinant, expressed in mammalian cells, ≥95% (SDS-PAGE), ≥95% (HPLC)
RAB0740 Human THBS1 / Thrombospondin-1 ELISA Kit, for serum, plasma, cell culture supernatants and urine
SAB4100184
Monoclonal Anti-BECN1 antibody produced in mouse, clone BECN1-B129, culture supernatant
SAB4100208
Monoclonal Anti-BECN1 antibody produced in mouse, clone BECN1-A129, culture supernatant
SAB5300513 Monoclonal Anti-BECN1 antibody produced in mouse, clone 2A4, ascites fluid
SAB1402138
Monoclonal Anti-CANX, (C-terminal) antibody produced in mouse, clone 1D4, purified immunoglobulin, buffered aqueous solution
SAB4700190 Monoclonal Anti-CD47 antibody produced in mouse, clone MEM-122, purified immunoglobulin, buffered aqueous solution
SAB1409170 Monoclonal Anti-GZMA antibody produced in mouse, clone 4C6, purified immunoglobulin, buffered aqueous solution
WH0027158M1
Monoclonal Anti-NDOR1 antibody produced in mouse, clone 3A11, purified immunoglobulin, buffered aqueous solution
SAB1405033
Monoclonal Anti-NDOR1 antibody produced in mouse, clone 3A11, ascites fluid
SAB4100604
Monoclonal Anti-NDOR1 antibody produced in mouse, clone NDOR1-A590, culture supernatant