EMAIL THIS PAGE TO A FRIEND
Carbohydrate polymers
Development of a rapid method for the quantification of cellulose in tobacco by (13)C CP/MAS NMR.
PMID 26453859
Abstract
A method was developed for rapid quantitative determination of cellulose in tobacco by utilizing (13)C cross polarization magic angle spinning NMR spectroscopy ((13)C CP/MAS NMR). Sample powder was loaded into NMR rotor, which was customized rotor containing a matched silicon tube as an intensity reference. (13)C CP/MAS NMR spectra of tobacco samples were processed with spectral deconvolution to obtain the area of the C-1 resonance at 105.5ppm and the internal standard at 0ppm. The ratio between the area of 105.5ppm and 0ppm of a set of standard cellulose samples was used to construct a calibration curve. The cellulose content of a tobacco sample was determined by comparison of the ratio between the area of 105.5ppm and 0ppm to the calibration curve. Results of this developed method showed good agreement with those obtained from chemical analysis. The proposed method has such advantages of accuracy, quickness and efficiency, and could be an alternative to chemical analyses of cellulose.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
718203
Allyl methyl sulfone, 96%
C4H8O2S