EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Identification of putative odorant binding protein genes in Asecodes hispinarum, a parasitoid of coconut leaf beetle (Brontispa longissima) by antennal RNA-Seq analysis.
PMID 26454175
Abstract
Asecodes hispinarum (Hymenoptera: Eulophidae) is an endoparasitoid and an efficient biological control agent which attacks larvae of Brontispa longissima, a serious insect pest of Palmae plants in China. Odorant binding proteins (OBPs) are believed to be important for transporting semiochemicals through the aqueous sensillar lymph to the olfactory receptor cells within the insect antennal sensilla. No previous study has reported on OBPs in A.xa0hispinarum. In this study, we conducted the large-scale identification of OBP genes from the antennae of A.xa0hispinarum by using transcriptome sequencing. Approximately 28.4 million total raw reads and about 27.3 million total clean reads were obtained, and then 46,363 unigenes were assembled. Of these unigenes, a total of 21,263 can be annotated in the NCBI non-redundant database. Among the annotated unigenes, 16,623 of them can be assigned to GO (Gene Ontology). Furthermore, we identified 8 putative OBP genes, and a phylogenetic tree analysis was performed to characterize the 8 OBP genes. In addition, the expression of the 8 OBP genes in different A.xa0hispinarum body tissues was analyzed by real-time quantitative polymerase chain reaction (qRT-PCR). The results indicated that the 8 OBP genes were expressed accordingly to sexes and tissues, but all highly expressed in antennae. The finding of this study will lay the foundation for unraveling molecular mechanisms of A.xa0hispinarum chemoperception.