EMAIL THIS PAGE TO A FRIEND
Cancer letters
Marmesin is a novel angiogenesis inhibitor: Regulatory effect and molecular mechanism on endothelial cell fate and angiogenesis.
PMID 26455771
Abstract
In the present study, we investigated the effects and molecular mechanism of marmesin, a coumarin compound isolated from Broussonetia kazinoki, on vascular endothelial growth factor-A (VEGF-A)-induced endothelial cell responses in vitro and angiogenic sprouting in aortic rings ex vivo. Marmesin treatment inhibited VEGF-A-stimulated endothelial cell proliferation through down-regulation of cell cycle-related proteins including cyclin-dependent kinases and cyclins, leading to pRb hypophosphorylation and G1 phase cell cycle arrest. In addition, marmesin treatment abrogated VEGF-A-induced endothelial cell migration, invasion and capillary-like structure formation in vitro as well as angiogenic sprouting ex vivo. These anti-angiogenic activities of marmesin were mediated through inactivation of VEGF-A-stimulated signaling pathways, and down-regulation of cell surface signaling molecules including VEGF receptor-2, human epidermal growth factor receptor-2, integrin β1 and integrin-liked kinase. Taken together, these findings clearly support the pharmacological roles of marmesin in regulating angiogenesis, and warrant further evaluation and development as a potential therapeutic agent for the treatment and prevention of angiogenesis-related diseases including cancer.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1400043
Anti-CCNE1 antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
HPA018169
Anti-CCNE1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1400044
Anti-CCNE1 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
AV01008 Anti-CDK2 (AB1) antibody produced in rabbit, IgG fraction of antiserum
AV03006
Anti-CDK2 (AB2) antibody produced in rabbit, IgG fraction of antiserum
C5223 Anti-Cdk2 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous glycerol solution
SAB4503706
Anti-CDK2 antibody produced in rabbit, affinity isolated antibody
SAB4500052
Anti-CDK2, C-Terminal antibody produced in rabbit, affinity isolated antibody
AV00001
Anti-CDK4 (AB1) antibody produced in rabbit, IgG fraction of antiserum
AV03017
Anti-CDK4 (AB2) antibody produced in rabbit, IgG fraction of antiserum
GW22309F Anti-CDK4 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1405599
Anti-CDK4 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB4300695
Anti-CDK4 antibody produced in rabbit, affinity isolated antibody
SAB4503515
Anti-Cyclin E1 antibody produced in rabbit, affinity isolated antibody
SAB4503516
Anti-Cyclin E1 antibody produced in rabbit, affinity isolated antibody
SAB4503514
Anti-Cyclin E1, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4300665
Anti-FAK (Ab-397) antibody produced in rabbit, affinity isolated antibody
SAB4502495
Anti-FAK antibody produced in rabbit, affinity isolated antibody
SAB4502496
Anti-FAK antibody produced in rabbit, affinity isolated antibody
SAB4502498
Anti-FAK antibody produced in rabbit, affinity isolated antibody
SAB4502499
Anti-FAK antibody produced in rabbit, affinity isolated antibody
SAB4502503
Anti-FAK antibody produced in rabbit, affinity isolated antibody
SAB4502497
Anti-FAK, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4502500
Anti-FAK, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4502501
Anti-FAK, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4502502
Anti-FAK, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4500784
Anti-HER2, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4500785
Anti-HER2, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4500786
Anti-HER2, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4500788
Anti-HER2, C-Terminal antibody produced in rabbit, affinity isolated antibody