EMAIL THIS PAGE TO A FRIEND
ACS nano
Polymer-Grafted Mesoporous Silica Nanoparticles as Ultrasound-Responsive Drug Carriers.
PMID 26456489
Abstract
A new ultrasound-responsive system based on mesoporous silica nanoparticles was developed for biomedical applications, grafting a copolymer on their surface that acts as gatekeeper of the pores. The nanoparticles can be loaded with a cargo at low temperature (4 °C), taking advantage of the open conformation that the polymer presents under these conditions. Then, at 37 °C the copolymer collapses closing the pore entrances and allowing the nanoparticles to carry the drugs at physiological temperature without premature release, which is of great importance when dealing with cytotoxic drugs in cancer treatments. Upon ultrasound irradiation, the sensitive polymer changes its hydrophobicity and, therefore, its conformation toward coil-like opening the gates and releasing the cargo. These hybrid nanoparticles have been shown to be noncytotoxic and can be internalized into LNCaP cells retaining their ultrasound-responsive capability in the cytoplasm of the cells. Moreover, doxorubicin-loaded hybrid MSNs were incubated with LNCaP cells to show their capacity to induce cell death only when the nanoparticles had been exposed to ultrasound. This work demonstrates that our hybrid-MSNs can be triggered by remote stimuli, which is of capital importance for future applications in drug delivery and cancer therapy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D106208
3,4-Dihydro-2H-pyran, 97%
C5H8O
11590
4,4′-Azobis(4-cyanovaleric acid), ≥98.0% (T)
C12H16N4O4
118168
4,4′-Azobis(4-cyanovaleric acid), ≥75%
C12H16N4O4
A3795
Ammonium nitrate, plant cell culture tested
H4N2O3
A9642
Ammonium nitrate, ≥99.0%
H4N2O3
256064
Ammonium nitrate, 99.999% trace metals basis
H4N2O3
221244
Ammonium nitrate, ACS reagent, ≥98%
H4N2O3
A7455
Ammonium nitrate, BioXtra, ≥99.5%
H4N2O3
31114
Ammonium nitrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥98%
H4N2O3
804819
Ammonium nitrate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥98%
H4N2O3
379115
DCC, 1.0 M in methylene chloride
C13H22N2
D80002
DCC, 99%
C13H22N2
447927
Di(ethylene glycol) methyl ether methacrylate, 95%
C9H16O4
F6377
Fluorescein sodium salt, used as fluorescent tracer
C20H10Na2O5
46960
Fluorescein sodium salt, BioReagent, suitable for fluorescence
C20H10Na2O5
46970
Fluorescein sodium salt, p.a., for source staining
C20H10Na2O5
155721
Methacrylic acid, contains 250 ppm MEHQ as inhibitor, 99%
C4H6O2
688886
Methyl acetoacetate, Lonza quality, ≥99% (GC)
C5H8O3
10913
Methyl acetoacetate, produced by Wacker Chemie AG, Burghausen, Germany, ≥99.0% (GC)
C5H8O3
537365
Methyl acetoacetate, ReagentPlus®, 99%
C5H8O3
283924
Rhodamine B isothiocyanate, mixed isomers
C29H30ClN3O3S
R1755
Rhodamine B isothiocyanate, BioReagent, mixed isomers, suitable for protein labeling
C29H30ClN3O3S