EMAIL THIS PAGE TO A FRIEND
The journal of pain : official journal of the American Pain Society
Clinically Effective Treatment of Fibromyalgia Pain With High-Definition Transcranial Direct Current Stimulation: Phase II Open-Label Dose Optimization.
PMID 26456677
Abstract
Despite promising preliminary results in treating fibromyalgia (FM) pain, no neuromodulation technique has been adopted in clinical practice because of limited efficacy, low response rate, or poor tolerability. This phase II open-label trial aims to define a methodology for a clinically effective treatment of pain in FM by establishing treatment protocols and screening procedures to maximize efficacy and response rate. High-definition transcranial direct current stimulation (HD-tDCS) provides targeted subthreshold brain stimulation, combining tolerability with specificity. We aimed to establish the number of HD-tDCS sessions required to achieve a 50% FM pain reduction, and to characterize the biometrics of the response, including brain network activation pain scores of contact heat-evoked potentials. We report a clinically significant benefit of a 50% pain reduction in half (nxa0=xa07) of the patients (Nxa0=xa014), with responders and nonresponders alike benefiting from a cumulative effect of treatment, reflected in significant pain reduction (Pxa0=xa0.035) as well as improved quality of life (Pxa0=xa0.001) over time. We also report an aggregate 6-week response rate of 50% of patients and estimate 15 as the median number of HD-tDCS sessions to reach clinically meaningful outcomes. The methodology for a pivotal FM neuromodulation clinical trial with individualized treatment is thus supported. Registered in Clinicaltrials.gov under registry number NCT01842009. In this article, an optimized protocol for the treatment of fibromyalgia pain with targeted subthreshold brain stimulation using high-definition transcranial direct current stimulation is outlined.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L512087
PPT, AldrichCPR
C36H26O2P2S