EMAIL THIS PAGE TO A FRIEND
ChemSusChem
In Situ-Grown ZnCo2O4 on Single-Walled Carbon Nanotubes as Air Electrode Materials for Rechargeable Lithium-Oxygen Batteries.
PMID 26457378
Abstract
The development of highly efficient catalysts is critical for the practical application of lithium-oxygen (Li-O2) batteries. Nanosheet-assembled ZnCo2O4 (ZCO) microspheres and thin films grown in situ on single-walled carbon nanotube (ZCO/SWCNT) composites as high-performance air electrode materials for Li-O2 batteries are reported. The in situ grown ZCO/SWCNT electrodes delivered high discharge capacities, decreased the onset of the oxygen evolution reaction by 0.9 V during the charging process, and led to longer cycling stability. These results indicate that in situ grown ZCO/SWCNT composites can be used as highly efficient air electrode materials for oxygen reduction and evolution reactions. The enhanced catalytic activity displayed by the uniformly dispersed ZCO catalyst on nanostructured electrodes is expected to inspire further development of other catalyzed electrodes for Li-O2 batteries and other applications.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
203106
Cobalt(II) nitrate hexahydrate, 99.999% trace metals basis
CoN2O6 · 6H2O
239267
Cobalt(II) nitrate hexahydrate, ACS reagent, ≥98%
CoN2O6 · 6H2O
230375
Cobalt(II) nitrate hexahydrate, reagent grade, 98%
CoN2O6 · 6H2O
60832
Cobalt(II) nitrate hexahydrate, puriss. p.a., ACS reagent, Ni ≤0.001%, ≥99.0% (KT)
CoN2O6 · 6H2O
499811
Lithium, granular, 99% trace metals basis
Li
444456
Lithium, granular, 4-10 mesh particle size, high sodium, 99% (metals basis)
Li
265977
Lithium, ingot, diam. 5.7 cm, 99.9% trace metals basis
Li
266000
Lithium, ribbon, thickness × W 1.5 mm × 100 mm, 99.9% trace metals basis
Li
265993
Lithium, ribbon, thickness × W 0.75 mm × 45 mm, 99.9% trace metals basis
Li
320080
Lithium, ribbon, thickness × W 0.75 mm × 19 mm, 99.9% trace metals basis
Li
265985
Lithium, ribbon, thickness × W 0.38 mm × 23 mm, 99.9% trace metals basis
Li
265969
Lithium, rod, diam. 12.7 mm, 99.9% trace metals basis
Li
220914
Lithium, wire (in mineral oil), diam. 3.2 mm, 99.9% trace metals basis
Li
278327
Lithium, wire, diam. 3.2 mm, in mineral oil, ≥98%
Li
GF47159826
Lithium, foil, 25x100mm, thickness 0.6mm, as rolled, 99.9%
Li
GF11639628
Lithium, foil, not light tested, 45x200mm, thickness 0.12mm, as rolled, 99.9%
Li
GF40266682
Lithium, foil, not light tested, 38x500mm, thickness 0.20mm, as rolled, 99.9%
Li
GF62158596
Lithium, foil, not light tested, 38x200mm, thickness 0.20mm, as rolled, 99.9%
Li
481548
Lithium trifluoromethanesulfonate, 99.995% trace metals basis
CF3LiO3S
282669
Lithium trifluoromethanesulfonate, 96%
CF3LiO3S
601535
Lithium-7Li, ≥98 atom % 7Li, ≥98% (CP)
7Li
172405
Tetraethylene glycol dimethyl ether, ≥99%
C10H22O5