EMAIL THIS PAGE TO A FRIEND
Neurogastroenterology and motility : the official journal of the European Gastrointestinal Motility Society
Acupuncture at heterotopic acupoints facilitates distal colonic motility via activating M3 receptors and somatic afferent C-fibers in normal, constipated, or diarrhoeic rats.
PMID 26459908
Abstract
Previous studies have demonstrated the efficacy of somatic stimulation for patients with gastrointestinal motility disorders. However, little effort has been made to investigate the effects of acupuncture on colonic motility, particularly in pathological conditions. The precise mechanism employed in the regulation of acupuncture on colonic motility still remains unclear. We assessed the effect of acupuncture at heterotopic acupoints on distal colonic motility using a warm-water-filled manometric balloon inserted 5-6 cm into the rectum of anesthetized normal rats or rats with diarrhea or constipation. Choline chloride, 4-DAMP, cobra venom and capsaicin were separately applied to investigate the role of M3 receptors in the regulation of distal colonic motility by acupuncture at heterotopic acupoints, and whether Aδ- and/or C-fibers are required for triggering distal colonic motility by acupuncture. Acupuncture at heterotopic acupoints increased distal colonic motility not only in normal rats but also in rats with constipation or diarrhea. M3 receptors play an important role in the facilitation of distal colonic motility triggered by acupuncture at heterotopic acupoints. Afferent nerve Aδ- and C-fibers mediate the transduction of the acupuncture signal and C-fibers are essential for enhancing the effect of acupuncture at the heterotopic acupoint on distal colonic motility. Our results reveal that acupuncture at heterotopic acupoints increases distal colonic motility regardless of normal or pathological conditions via predominately activating C-fibers of somatic afferent nerve and M3 receptors.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0255
4-DAMP, ≥98% (HPLC)
C21H26INO2