EMAIL THIS PAGE TO A FRIEND
Biologicals : journal of the International Association of Biological Standardization
Reverse vaccinology as an approach for developing Histophilus somni vaccine candidates.
PMID 26460173
Abstract
Histophilosis of cattle is caused by the Gram negative bacterial pathogen Histophilus somni (H. somni) which is also associated with the bovine respiratory disease (BRD) complex. Existing vaccines for H.xa0somni include either killed cells or bacteria-free outer membrane proteins from the organism which have proven to be moderately successful. In this study, reverse vaccinology was used to predict potential H.xa0somni vaccine candidates from genome sequences. In turn, these may protect animals against new strains circulating in the field. Whole genome sequencing of six recent clinical H.xa0somni isolates was performed using an Illumina MiSeq and compared to six genomes from the 1980's. De novo assembly of crude whole genomes was completed using Geneious 6.1.7. Protein coding regions was predicted using Glimmer3. Scores from multiple web-based programs were utilized to evaluate the antigenicity of these predicted proteins which were finally ranked based on their surface exposure scores. A single new strain was selected for future vaccine development based on conservation of the protein candidates among all 12 isolates. A positive signal with convalescent serum for these antigens in western blots indicates inxa0vivo recognition. In order to test the protective capacity of these antigens bovine animal trials are ongoing.