EMAIL THIS PAGE TO A FRIEND
Oncotarget
PRMT1 promotes mitosis of cancer cells through arginine methylation of INCENP.
PMID 26460953
Abstract
Inner centromere protein (INCENP) is a part of a protein complex known as the chromosomal passenger complex (CPC) that is essential for correcting non-bipolar chromosome attachments and for cytokinesis. We here demonstrate that a protein arginine methyltransferase PRMT1, which are overexpressed in various types of cancer including lung and bladder cancer, methylates arginine 887 in an Aurora Kinase B (AURKB)-binding region of INCENP both in vitro and in vivo. R887-substituted INCENP revealed lower binding-affinity to AURKB than wild-type INCENP in the presence of PRMT1. Knockdown of PRMT1 as well as overexpression of methylation-inactive INCENP attenuated the AURKB activity in cancer cells, and resulted in abnormal chromosomal alignment and segregation. Furthermore, introduction of methylation-inactive INCENP into cancer cells reduced the growth rate, compared with those introduced wild-type INCENP or Mock. Our data unveils a novel mechanism of PRMT1-mediated CPC regulation through methylation of INCENP.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A5102 Anti-Aurora B antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB4500348
Anti-Histone H3 (Acetyl-Lys14) antibody produced in rabbit, affinity isolated antibody
SAB4500349
Anti-Histone H3 (Acetyl-Lys18) antibody produced in rabbit, affinity isolated antibody
SAB4500350
Anti-Histone H3 (Acetyl-Lys23) antibody produced in rabbit, affinity isolated antibody
SAB4500351
Anti-Histone H3 (Acetyl-Lys27) antibody produced in rabbit, affinity isolated antibody
SAB4500347
Anti-Histone H3 (Acetyl-Lys9) antibody produced in rabbit, affinity isolated antibody
H9289
Anti-Histone H3 (N-terminal) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
H0164
Anti-Histone H3 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
I5283 Anti-INCENP antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB4504219
Anti-phospho-AurB (pThr232) antibody produced in rabbit, affinity isolated antibody
SAB4504274
Anti-phospho-AurB (pTyr12) antibody produced in rabbit, affinity isolated antibody
H0412 Anti-phospho-Histone H3 (pSer10) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB4503877
Anti-phospho-Histone H3 (pSer28) antibody produced in rabbit, affinity isolated antibody
SAB4503876
Anti-phospho-Histone H3 (pThr3) antibody produced in rabbit, affinity isolated antibody
P6996
Anti-PRMT1 (TK-16) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
P6871 Anti-PRMT1 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
P1620
Anti-PRMT1 antibody, Mouse monoclonal, clone PRMT1-171, purified from hybridoma cell culture
S5386
DL-Serine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)
C3H7NO3
S4375
DL-Serine, ≥98% (TLC)
C3H7NO3
27002
Formic acid, purum, ≥85%
CH2O2
WH0009212M3
Monoclonal Anti-AURKB antibody produced in mouse, clone 6H7, purified immunoglobulin, buffered aqueous solution
SAB5300517
Monoclonal Anti-AURKB antibody produced in mouse, clone 13E8D3, ascites fluid
SAB1404685
Monoclonal Anti-AURKB, (N-terminal) antibody produced in mouse, clone 6G8, purified immunoglobulin, buffered aqueous solution
H9908
Monoclonal Anti-phospho-Histone H3 (pSer28) antibody produced in rat, ~0.5 mg/mL, clone HTA28, purified immunoglobulin, buffered aqueous solution
SRP0141
PRMT1 Active human, recombinant, expressed in baculovirus infected insect cells, ≥70% (SDS-PAGE)
P0083 PRMT1 from rat, recombinant, expressed in E. coli, ≥90% (SDS-PAGE), buffered aqueous solution
SRP0140
PRMT1 human, recombinant, expressed in E. coli, ≥50% (SDS-PAGE)
PLA0148
Rabbit anti-Histone H3 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.