EMAIL THIS PAGE TO A FRIEND
EMBO reports
From ribosomes to royal charters: An interview with Venkatraman Ramakrishnan from the LMB in Cambridge, UK, and President Elect of the Royal Society.
PMID 26464472