EMAIL THIS PAGE TO A FRIEND
International journal of clinical and experimental pathology
Overexpression of metabolic markers HK1 and PKM2 contributes to lymphatic metastasis and adverse prognosis in Chinese gastric cancer.
PMID 26464675
Abstract
Hexokinase 1 (HK1) and pyruvate kinase M2 (PKM2) are two key regulators in glycosis and oncogenic markers in cancers. In the present study, we investigated the expression profile by Western blotting and immunohistochemistry and determined their prognostic values in the gastric cancer. Expression of HK1 and PKM2 was remarkably increased in gastric cancer tissues and was significantly associated lymphatic metastasis and advanced TNM staging. In the COX regression model, HK1 and TNM stage were analyzed as adverse prognostic indicators in gastric cancer. Furthermore, patients with HK1 expression showed remarkable shorter survival duration in both lymphatic metastasis cohort and advanced staging cohort. Our results suggest that overexpression of PKM2 and HK1, especially the latter, significantly associates with lymphatic metastasis, advanced clinical staging and unfavorable prognosis in gastric cancer.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1409191
Anti-HK1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2701748
Anti-hk1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA007044
Anti-HK1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA007043
Anti-HK1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410630
Anti-HK1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1407479
Anti-HOOK1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA018537
Anti-HOOK1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
HPA018820
Anti-HOOK1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
SAB1410920
Anti-HOOK1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA016422
Anti-KCNA4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1302037
ANTI-TAC4 (C-TERM) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
WH0003098M1
Monoclonal Anti-HK1 antibody produced in mouse, clone 5G9, purified immunoglobulin, buffered aqueous solution
SAB5300279 Monoclonal Anti-HK1 antibody produced in mouse, clone 7A7, ascites fluid
SAB5300280 Monoclonal Anti-HK1 antibody produced in mouse, clone 3A10, ascites fluid
SAB1400552
Monoclonal Anti-HOOK1 antibody produced in mouse, clone 1D11, purified immunoglobulin, buffered aqueous solution
SAB5200058
Monoclonal Anti-KCNA4 antibody produced in mouse, clone S13-31, 1 mg/mL, purified immunoglobulin