EMAIL THIS PAGE TO A FRIEND
Investigative ophthalmology & visual science
Application of Graphene as Candidate Biomaterial for Synthetic Keratoprosthesis Skirt.
PMID 26465888
Abstract
Synthetic keratoprostheses are required for visual rehabilitation in patients with end-stage corneal blindness. This study aimed to assess the biocompatibility of graphene material and its potential as a novel synthetic keratoprosthesis skirt material for corneal tissue engineering. Human corneal stromal fibroblasts were cultured on material surfaces including pristine graphene film, graphene foam, pristine titanium (Ti) discs, and tissue culture plastic surface (TCPS). Cell attachment was assayed by immunostaining of paxillin and vinculin. Cell viability and proliferation were assessed by MTT (3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide) and Click iT 5-ethynyl-2'-deoxyuridine (EdU) assays. The growth of fibroblasts on three-dimensional graphene foam was examined by scanning electron microscopy, and cytokine release was analyzed by enzyme-linked immunosorbent assay. Graphene films were implanted into rabbit corneal stromal pockets and examined by slit-lamp biomicroscopy, anterior segment optical coherence tomography, in vivo confocal microscopy, and histology. Pristine graphene demonstrated good biocompatibility with human stromal fibroblasts in terms of cell adhesion, viability, and proliferation. Cells on graphene films showed higher number than on TCPS control. Cells grown on graphene had 10% more proliferation than on Ti. The expression levels of IL-6 and IL-8 were reduced when cells were seeded on graphene foam as compared to Ti and graphene film. Implantation of graphene film into rabbit stroma (n = 6) did not show any signs of infection, neovascularization, or inflammation. Graphene displayed excellent short-term biocompatibility with corneal cells and tissue. This demonstrates that graphene can be developed as a tissue engineering material for use in cornea.