EMAIL THIS PAGE TO A FRIEND
Journal of immunology (Baltimore, Md. : 1950)
Cytosolic Low Molecular Weight FGF2 Orchestrates RIG-I-Mediated Innate Immune Response.
PMID 26466960
Abstract
Fibroblast growth factor (FGF)2,which is one of the 22 members of the FGF family, functions as an extracellular molecule involved in canonical receptor tyrosine kinase signaling. It has been implicated in angiogenesis and the development of the CNS. In this article, we reveal that cytosolic low m.w. isoform (LMW) FGF2 (18 kDa), not its secreted form, plays an unexpected role in the innate immune response. Cytosolic LMW FGF2 directly associated with inactivated RIG-I under physiological conditions, which enhanced RIG-I protein stability, thereby maintaining basal RIG-I levels. However, during RIG-I activation induced by viral RNA, cytosolic FGF2 bound to the caspase recruitment domains of activated RIG-I, which blocked RIG-I-MAVS complex formation. LMW FGF2 deficiency increased type I IFN production, whereas the overexpression of LMW FGF2 exerted the opposite effect. Cytosolic LMW FGF2 functions as a negative regulator in RIG-I-mediated antiviral signaling. This work provides insight into the role of FGF2 in innate immune response.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1400461
Anti-DDX58 antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
SAB2104315
Anti-DDX58 antibody produced in rabbit, affinity isolated antibody
SAB1407860
Anti-IFIH1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA002656
Anti-IFIH1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4501565
Anti-IRF-3, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB1406043
Anti-IRF3 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA004895
Anti-IRF3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV31992
Anti-IRF3 antibody produced in rabbit, affinity isolated antibody, lyophilized powder
AV38061
Anti-IRF3 antibody produced in rabbit, IgG fraction of antiserum, lyophilized powder
SAB3500280
Anti-IRF3 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4501563
Anti-IRF3, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4501564
Anti-IRF3, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB3500356
Anti-MDA5 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4503926
Anti-phospho-IRF-3 (pSer385) antibody produced in rabbit, affinity isolated antibody
SAB4504651
Anti-phospho-IRF-3 (pSer386) antibody produced in rabbit, affinity isolated antibody
SAB4504031
Anti-phospho-IRF-3 (pSer396) antibody produced in rabbit, affinity isolated antibody
PRS3953
Anti-RIG-1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1300230 Anti-RIG-I (C-term I790) antibody produced in rabbit, saturated ammonium sulfate (SAS) precipitated, buffered aqueous solution
SAB1300062 Anti-RIG-I (C-term) antibody produced in rabbit, saturated ammonium sulfate (SAS) precipitated, buffered aqueous solution
SAB1300231
Anti-RIG-I (center) antibody produced in rabbit, saturated ammonium sulfate (SAS) precipitated, buffered aqueous solution
SAB1305862
ANTI-RIG-I (CENTER) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1305861 ANTI-RIG-I(C-TERMINAL) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1402619
Monoclonal Anti-IFIH1 antibody produced in mouse, clone 3F6, purified immunoglobulin, buffered aqueous solution
SAB4100036
Monoclonal Anti-IRF3 antibody produced in mouse, culture supernatant
SAB4100037
Monoclonal Anti-IRF3 antibody produced in mouse, culture supernatant
SAB1402824
Monoclonal Anti-IRF3 antibody produced in mouse, clone 3C8, purified immunoglobulin, buffered aqueous solution