EMAIL THIS PAGE TO A FRIEND
Food chemistry
Effect of carob (Ceratonia siliqua L.) flour on the antioxidant potential, nutritional quality, and sensory characteristics of fortified durum wheat pasta.
PMID 26471602
Abstract
This paper presents a study on the effect of carob flour addition from 1% to 5% (w/w) on phenolics content, antioxidant activity, nutritional quality, and sensory attributes of wheat pasta. An increase of about 2-folds, 18-folds and 3-folds in phenolics content, antiradical activity and reducing power for pasta fortified with 5% of carob flour was observed, respectively, compared to the control. Expected glycemic index (eGI) was increased proportionally to the substitution level and ranged between 72.2 and 83.9 for 1-5% of supplement, respectively. Furthermore, pasta fortification affected the in vitro bioaccessibility of nutrients. In case of 5% supplemented pasta, the digestibility of starch and protein decreased by about 9% compared to the control. The replacement of semolina with carob flour from 1% to 5% had no significant effect on pasta sensory attributes. In conclusion, carob flour seems to be a promising functional ingredient for pasta fortification.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
218898
5-(Dimethylamino)-1-naphthalenesulfonamide, 99%
C12H14N2O2S