EMAIL THIS PAGE TO A FRIEND
Food chemistry
Cookies from composite wheat-sesame peels flours: dough quality and effect of Bacillus subtilis SPB1 biosurfactant addition.
PMID 26471616
Abstract
Sesame coat is a valuable by-product. The study was carried out on sesame peels flour at different replacing levels of white wheat flour in five cookies dough formulations. The functional properties of composite flours such as swelling capacity, water holding capacity, oil holding capacity, emulsifying capacity, foam capacity, gelatinization temperature, least gelation concentration and bulk density were increased with increase in the sesame peels flour incorporation along with wheat flour. Texture analysis of dough revealed that, the addition of sesame peels flour affected the quality of dough in terms of hardness, cohesion, adhesion and breaking strength. Cookies supplemented with sesame peels flour showed interesting physical properties with lower moisture content and higher spread factor than those made by white wheat flour. But, their hardness increase with the increase of the replacement ratio and their color becomes indesirable. Interestingly, sensory results indicated that cookies supplemented with sesame peels flour were acceptable at a level that not exceeds 30% of incorporation. By the addition of SPB1 biosurfactant at 0.1%, the dough texture profile was significantly improved and the action of this bioemulsifier was more pronounced than a commercial emulsifier known as glycerol monostearate. With the addition of SPB1 biosurfactant on cookies' dough, we manage to obtain cookies softer and with better overall quality.