EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Microchidia protein 2, MORC2, downregulates the cytoskeleton adapter protein, ArgBP2, via histone methylation in gastric cancer cells.
PMID 26476214
Abstract
ArgBP2 is an adapter protein that plays an important role in actin-dependent processes such as cell adhesion and migration. However, its function and regulation mechanisms in gastric cancer have not yet been investigated. Here, we showed the low expression of ArgBP2 mRNA level in gastric tumor samples and its repressive function in the proliferation, migration, and invasion of gastric cancer cells. Then, we cloned and identified ArgBP2 promoter and verified that MORC2 bound to the promoter. Moreover, we demonstrated that MORC2 enhanced the recruitment of EZH2, which promoted the tri-methylation of H3K27, leading to the transcriptional repression of ArgBP2. Our results might thus contribute to understanding the molecular mechanisms of ArgBP2 regulation and suggesting ArgBP2 as a potential therapeutic target for gastric cancer.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1409110
Anti-ABL2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1411998
ANTI-ABL2 antibody produced in mouse, clone 3G3, purified immunoglobulin, buffered aqueous solution
SAB1411999
ANTI-ABL2 antibody produced in mouse, clone 3D2, purified immunoglobulin, buffered aqueous solution
SAB1412281
ANTI-ABL2 antibody produced in mouse, clone 5C7, purified immunoglobulin, buffered aqueous solution
SAB1412323
ANTI-ABL2 antibody produced in mouse, clone 3E4, purified immunoglobulin, buffered aqueous solution
SAB1412446
ANTI-ABL2 antibody produced in mouse, clone 2H8, purified immunoglobulin, buffered aqueous solution
HPA001866
Anti-ABL2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA000436
Anti-MORC2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
WH0000027M9
Monoclonal Anti-ABL2 antibody produced in mouse, clone 5C6, purified immunoglobulin, buffered aqueous solution
SAB1403515
Monoclonal Anti-ABL2 antibody produced in mouse, clone 6D5, purified immunoglobulin, buffered aqueous solution
SAB5300521
Monoclonal Anti-ABL2 antibody produced in mouse, clone 1H1B11, ascites fluid
SAB5300522 Monoclonal Anti-ABL2 antibody produced in mouse, clone 1H1, ascites fluid