EMAIL THIS PAGE TO A FRIEND
Small (Weinheim an der Bergstrasse, Germany)
Monolayer Silane-Coated, Water-Soluble Quantum Dots.
PMID 26476710
Abstract
A one-step method to produce ≈12 nm hydrodynamic diameter water-soluble CdSe/ZnS quantum dots (QDs), as well as CdS/ZnS, ZnSe/ZnMnS/ZnS, AgInS2 /ZnS, and CuInS2 /ZnS QDs, by ligand exchange with a near-monolayer of organosilane caps is reported. The method cross-links the surface-bound silane ligands such that the samples are stable on the order of months under ambient conditions. Furthermore, the samples may retain a high quantum yield (60%) over this time. Several methods to functionalize aqueous QD dispersions with proteins and fluorescent dyes have been developed with reaction yields as high as 97%.