EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
Evolution and structural organization of the mitochondrial contact site (MICOS) complex and the mitochondrial intermembrane space bridging (MIB) complex.
PMID 26477565
Abstract
We have analyzed the distribution of mitochondrial contact site and cristae organizing system (MICOS) complex proteins and mitochondrial intermembrane space bridging complex (MIB) proteins over (sub)complexes and over species. The MICOS proteins are associated with the formation and maintenance of mitochondrial cristae. Indeed, the presence of MICOS genes in genomes correlates well with the presence of cristae: all cristae containing species have at least one MICOS gene and cristae-less species have none. Mic10 is the most widespread MICOS gene, while Mic60 appears be the oldest one, as it originates in the ancestors of mitochondria, the proteobacteria. In proteobacteria the gene occurs in clusters with genes involved in heme synthesis while the protein has been observed in intracellular membranes of the alphaproteobacterium Rhodobacter sphaeroides. In contrast, Mic23 and Mic27 appear to be the youngest MICOS proteins, as they only occur in opisthokonts. The remaining MICOS proteins, Mic10, Mic19, Mic25 and Mic12, the latter we show to be orthologous to human C19orf70/QIL1, trace back to the root of the eukaryotes. Of the remaining MIB proteins, also DNAJC11 shows a high correlation with the presence of cristae. In mitochondrial protein complexome profiles, the MIB complex occurs as a defined complex and as separate subcomplexes, potentially reflecting various assembly stages. We find three main forms of the complex: A) The MICOS complex, containing all the MICOS proteins, B) a membrane bridging subcomplex, containing in addition SAMM50, MTX2 and the previously uncharacterized MTX3, and C) the complete MIB complex containing in addition DNAJC11 and MTX1.